Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAX1BP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_006024.6 | NP_006015.4 | 14 - 15 | 2123 | 96 | |
GenBank mRNA | AK000098.1 | - | 14 - 15 | 2036 | 96 | |
AK314292.1 | - | 14 - 15 | 2093 | 96 | ||
AL136586.1 | - | 14 - 15 | 2029 | 96 | ||
BC050358.2 | - | 14 - 15 | 2008 | 6055189 | 96 | |
CR533556.1 | - | 13 - 14 | 1941 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |