Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR07008362.hg.1 |
TC07000569.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07008362.hg.1 |
ENST00000257621.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07008362.hg.1 |
ENST00000325885.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07008362.hg.1 |
ENST00000428113.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07008362.hg.1 |
NM_016116.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07008362.hg.1 |
NM_145872.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008389.hg.1 |
ENST00000257621.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008389.hg.1 |
ENST00000257621.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008389.hg.1 |
ENST00000257621.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008389.hg.1 |
ENST00000325885.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008389.hg.1 |
ENST00000325885.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008389.hg.1 |
ENST00000325885.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008389.hg.1 |
ENST00000325885.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008389.hg.1 |
ENST00000325885.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008389.hg.1 |
ENST00000325885.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008389.hg.1 |
ENST00000325885.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008389.hg.1 |
ENST00000428113.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008389.hg.1 |
ENST00000428113.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008389.hg.1 |
ENST00000428113.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008389.hg.1 |
ENST00000428113.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008389.hg.1 |
ENST00000428113.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008389.hg.1 |
ENST00000428113.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008389.hg.1 |
ENST00000428113.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008389.hg.1 |
NM_016116.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008389.hg.1 |
NM_016116.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008389.hg.1 |
NM_016116.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008389.hg.1 |
NM_016116.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008389.hg.1 |
NM_016116.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008389.hg.1 |
NM_016116.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008389.hg.1 |
NM_016116.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008389.hg.1 |
NM_145872.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008389.hg.1 |
NM_145872.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008389.hg.1 |
NM_145872.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008389.hg.1 |
NM_145872.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008389.hg.1 |
NM_145872.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008389.hg.1 |
NM_145872.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008389.hg.1 |
NM_145872.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07008360.hg.1 |
TC07000569.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07008360.hg.1 |
ENST00000325885.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07008360.hg.1 |
ENST00000428113.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07008360.hg.1 |
NM_016116.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07008360.hg.1 |
NM_145872.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07008361.hg.1 |
TC07000569.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07008361.hg.1 |
ENST00000325885.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07008361.hg.1 |
ENST00000428113.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07008361.hg.1 |
NM_016116.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07008361.hg.1 |
NM_145872.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700158601.hg.1 |
ENST00000325885.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700158601.hg.1 |
ENST00000428113.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700158601.hg.1 |
NM_016116.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700158601.hg.1 |
NM_145872.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700158601.hg.1 |
ENST00000325885.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700158601.hg.1 |
ENST00000428113.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700158601.hg.1 |
NM_016116.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700158601.hg.1 |
NM_145872.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700158601.hg.1 |
TC0700008389.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700158602.hg.1 |
ENST00000325885.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700158602.hg.1 |
ENST00000428113.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700158602.hg.1 |
NM_016116.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700158602.hg.1 |
NM_145872.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700158602.hg.1 |
ENST00000325885.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700158602.hg.1 |
ENST00000428113.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700158602.hg.1 |
NM_016116.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700158602.hg.1 |
NM_145872.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700158602.hg.1 |
TC0700008389.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0700008389.hg.1 |
ENST00000257621.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008389.hg.1 |
ENST00000257621.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008389.hg.1 |
ENST00000257621.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008389.hg.1 |
ENST00000325885.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008389.hg.1 |
ENST00000325885.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008389.hg.1 |
ENST00000325885.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008389.hg.1 |
ENST00000325885.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008389.hg.1 |
ENST00000325885.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008389.hg.1 |
ENST00000325885.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008389.hg.1 |
ENST00000325885.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008389.hg.1 |
ENST00000428113.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008389.hg.1 |
ENST00000428113.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008389.hg.1 |
ENST00000428113.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008389.hg.1 |
ENST00000428113.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008389.hg.1 |
ENST00000428113.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008389.hg.1 |
ENST00000428113.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008389.hg.1 |
ENST00000428113.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008389.hg.1 |
NM_016116.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008389.hg.1 |
NM_016116.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008389.hg.1 |
NM_016116.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008389.hg.1 |
NM_016116.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008389.hg.1 |
NM_016116.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008389.hg.1 |
NM_016116.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008389.hg.1 |
NM_016116.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008389.hg.1 |
NM_145872.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008389.hg.1 |
NM_145872.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008389.hg.1 |
NM_145872.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008389.hg.1 |
NM_145872.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008389.hg.1 |
NM_145872.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008389.hg.1 |
NM_145872.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008389.hg.1 |
NM_145872.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700158603.hg.1 |
ENST00000257621.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700158603.hg.1 |
ENST00000325885.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700158603.hg.1 |
ENST00000428113.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700158603.hg.1 |
NM_016116.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700158603.hg.1 |
NM_145872.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700158603.hg.1 |
ENST00000257621.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700158603.hg.1 |
ENST00000325885.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700158603.hg.1 |
ENST00000428113.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700158603.hg.1 |
NM_016116.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700158603.hg.1 |
NM_145872.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700158603.hg.1 |
TC0700008389.hg.1 |
Clariom D TC |