Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR13002585.hg.1 |
ENST00000298125.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13002585.hg.1 |
NM_052950.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13002585.hg.1 |
TC13000217.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007228.hg.1 |
ENST00000298125.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007228.hg.1 |
ENST00000298125.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007228.hg.1 |
ENST00000298125.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007228.hg.1 |
ENST00000298125.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007228.hg.1 |
ENST00000298125.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007228.hg.1 |
ENST00000298125.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007228.hg.1 |
ENST00000298125.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007228.hg.1 |
ENST00000298125.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007228.hg.1 |
ENST00000298125.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007228.hg.1 |
ENST00000298125.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007228.hg.1 |
ENST00000298125.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007228.hg.1 |
ENST00000298125.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007228.hg.1 |
ENST00000298125.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007228.hg.1 |
ENST00000298125.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007228.hg.1 |
ENST00000298125.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007228.hg.1 |
ENST00000298125.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007228.hg.1 |
ENST00000298125.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007228.hg.1 |
ENST00000460145.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007228.hg.1 |
ENST00000460145.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007228.hg.1 |
ENST00000460145.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007228.hg.1 |
ENST00000612477.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007228.hg.1 |
ENST00000612477.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007228.hg.1 |
ENST00000612477.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007228.hg.1 |
ENST00000612477.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007228.hg.1 |
NM_052950.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007228.hg.1 |
NM_052950.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007228.hg.1 |
NM_052950.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007228.hg.1 |
NM_052950.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007228.hg.1 |
NM_052950.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007228.hg.1 |
NM_052950.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007228.hg.1 |
NM_052950.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007228.hg.1 |
NM_052950.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007228.hg.1 |
NM_052950.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007228.hg.1 |
NM_052950.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007228.hg.1 |
NM_052950.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007228.hg.1 |
NM_052950.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007228.hg.1 |
NM_052950.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007228.hg.1 |
NM_052950.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007228.hg.1 |
NM_052950.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007228.hg.1 |
NM_052950.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007228.hg.1 |
NM_052950.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007228.hg.1 |
NM_052950.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007228.hg.1 |
ENST00000298125.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007228.hg.1 |
ENST00000298125.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007228.hg.1 |
ENST00000298125.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007228.hg.1 |
ENST00000298125.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007228.hg.1 |
ENST00000298125.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007228.hg.1 |
ENST00000298125.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007228.hg.1 |
ENST00000298125.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007228.hg.1 |
ENST00000298125.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007228.hg.1 |
ENST00000298125.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007228.hg.1 |
ENST00000298125.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007228.hg.1 |
ENST00000298125.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007228.hg.1 |
ENST00000298125.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007228.hg.1 |
ENST00000298125.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007228.hg.1 |
ENST00000298125.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007228.hg.1 |
ENST00000298125.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007228.hg.1 |
ENST00000298125.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007228.hg.1 |
ENST00000298125.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007228.hg.1 |
ENST00000460145.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007228.hg.1 |
ENST00000460145.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007228.hg.1 |
ENST00000460145.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007228.hg.1 |
ENST00000612477.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007228.hg.1 |
ENST00000612477.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007228.hg.1 |
ENST00000612477.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007228.hg.1 |
ENST00000612477.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007228.hg.1 |
NM_052950.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007228.hg.1 |
NM_052950.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007228.hg.1 |
NM_052950.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007228.hg.1 |
NM_052950.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007228.hg.1 |
NM_052950.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007228.hg.1 |
NM_052950.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007228.hg.1 |
NM_052950.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007228.hg.1 |
NM_052950.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007228.hg.1 |
NM_052950.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007228.hg.1 |
NM_052950.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007228.hg.1 |
NM_052950.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007228.hg.1 |
NM_052950.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007228.hg.1 |
NM_052950.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007228.hg.1 |
NM_052950.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007228.hg.1 |
NM_052950.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007228.hg.1 |
NM_052950.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007228.hg.1 |
NM_052950.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007228.hg.1 |
NM_052950.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300149043.hg.1 |
ENST00000298125.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300149043.hg.1 |
NM_052950.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300149043.hg.1 |
ENST00000298125.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300149043.hg.1 |
NM_052950.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300149043.hg.1 |
TC1300007228.hg.1 |
Clariom D TC |