Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_000759.3 | NP_000750.1 | 2 - 3 | 317 | 92 | |
GenBank mRNA | X03438.1 | - | 2 - 3 | 234 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Probe spans exons |