Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR11018293.hg.1 |
TC11003476.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11018293.hg.1 |
ENST00000265970.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11018293.hg.1 |
ENST00000544896.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11018293.hg.1 |
NM_002645.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11018294.hg.1 |
TC11003476.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11018294.hg.1 |
ENST00000544896.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11018294.hg.1 |
NM_002645.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013148.hg.1 |
ENST00000265970.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013148.hg.1 |
ENST00000265970.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013148.hg.1 |
ENST00000265970.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013148.hg.1 |
ENST00000265970.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013148.hg.1 |
ENST00000265970.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013148.hg.1 |
ENST00000265970.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013148.hg.1 |
ENST00000265970.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013148.hg.1 |
ENST00000265970.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013148.hg.1 |
ENST00000265970.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013148.hg.1 |
ENST00000265970.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013148.hg.1 |
ENST00000265970.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013148.hg.1 |
ENST00000265970.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013148.hg.1 |
ENST00000265970.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013148.hg.1 |
ENST00000265970.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013148.hg.1 |
ENST00000265970.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013148.hg.1 |
ENST00000265970.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013148.hg.1 |
ENST00000265970.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013148.hg.1 |
ENST00000265970.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013148.hg.1 |
ENST00000265970.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013148.hg.1 |
ENST00000265970.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013148.hg.1 |
ENST00000265970.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013148.hg.1 |
ENST00000265970.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013148.hg.1 |
ENST00000265970.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013148.hg.1 |
ENST00000265970.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013148.hg.1 |
ENST00000265970.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013148.hg.1 |
ENST00000265970.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013148.hg.1 |
ENST00000265970.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013148.hg.1 |
ENST00000265970.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013148.hg.1 |
ENST00000265970.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013148.hg.1 |
ENST00000265970.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013148.hg.1 |
ENST00000265970.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013148.hg.1 |
ENST00000265970.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013148.hg.1 |
ENST00000265970.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013148.hg.1 |
ENST00000265970.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013148.hg.1 |
ENST00000265970.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013148.hg.1 |
ENST00000503729.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013148.hg.1 |
ENST00000503729.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013148.hg.1 |
ENST00000503729.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013148.hg.1 |
ENST00000532035.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013148.hg.1 |
ENST00000532035.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013148.hg.1 |
ENST00000532035.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013148.hg.1 |
ENST00000533645.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013148.hg.1 |
ENST00000533645.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013148.hg.1 |
ENST00000533645.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013148.hg.1 |
ENST00000533645.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013148.hg.1 |
ENST00000533645.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013148.hg.1 |
ENST00000533645.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013148.hg.1 |
ENST00000533645.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013148.hg.1 |
ENST00000533645.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013148.hg.1 |
ENST00000533645.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013148.hg.1 |
NM_002645.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013148.hg.1 |
NM_002645.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013148.hg.1 |
NM_002645.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013148.hg.1 |
NM_002645.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013148.hg.1 |
NM_002645.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013148.hg.1 |
NM_002645.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013148.hg.1 |
NM_002645.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013148.hg.1 |
NM_002645.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013148.hg.1 |
NM_002645.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013148.hg.1 |
NM_002645.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013148.hg.1 |
NM_002645.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013148.hg.1 |
NM_002645.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013148.hg.1 |
NM_002645.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013148.hg.1 |
NM_002645.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013148.hg.1 |
NM_002645.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013148.hg.1 |
NM_002645.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013148.hg.1 |
NM_002645.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013148.hg.1 |
NM_002645.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013148.hg.1 |
NM_002645.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013148.hg.1 |
NM_002645.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013148.hg.1 |
NM_002645.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013148.hg.1 |
NM_002645.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013148.hg.1 |
NM_002645.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013148.hg.1 |
NM_002645.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013148.hg.1 |
NM_002645.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013148.hg.1 |
NM_002645.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013148.hg.1 |
NM_002645.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013148.hg.1 |
NM_002645.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013148.hg.1 |
NM_002645.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013148.hg.1 |
NM_002645.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013148.hg.1 |
NM_002645.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013148.hg.1 |
NM_002645.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013148.hg.1 |
NM_002645.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013148.hg.1 |
NM_002645.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013148.hg.1 |
NM_002645.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013148.hg.1 |
NM_002645.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100203741.hg.1 |
ENST00000265970.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100203741.hg.1 |
ENST00000533645.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100203741.hg.1 |
NM_002645.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100203741.hg.1 |
ENST00000265970.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100203741.hg.1 |
ENST00000533645.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100203741.hg.1 |
NM_002645.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100203741.hg.1 |
TC1100013148.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100203743.hg.1 |
ENST00000532035.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100203743.hg.1 |
ENST00000533645.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100203743.hg.1 |
NM_002645.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100203743.hg.1 |
ENST00000532035.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100203743.hg.1 |
ENST00000533645.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100203743.hg.1 |
NM_002645.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100203743.hg.1 |
TC1100013148.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100013148.hg.1 |
ENST00000265970.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013148.hg.1 |
ENST00000265970.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013148.hg.1 |
ENST00000265970.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013148.hg.1 |
ENST00000265970.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013148.hg.1 |
ENST00000265970.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013148.hg.1 |
ENST00000265970.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013148.hg.1 |
ENST00000265970.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013148.hg.1 |
ENST00000265970.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013148.hg.1 |
ENST00000265970.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013148.hg.1 |
ENST00000265970.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013148.hg.1 |
ENST00000265970.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013148.hg.1 |
ENST00000265970.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013148.hg.1 |
ENST00000265970.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013148.hg.1 |
ENST00000265970.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013148.hg.1 |
ENST00000265970.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013148.hg.1 |
ENST00000265970.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013148.hg.1 |
ENST00000265970.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013148.hg.1 |
ENST00000265970.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013148.hg.1 |
ENST00000265970.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013148.hg.1 |
ENST00000265970.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013148.hg.1 |
ENST00000265970.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013148.hg.1 |
ENST00000265970.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013148.hg.1 |
ENST00000265970.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013148.hg.1 |
ENST00000265970.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013148.hg.1 |
ENST00000265970.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013148.hg.1 |
ENST00000265970.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013148.hg.1 |
ENST00000265970.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013148.hg.1 |
ENST00000265970.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013148.hg.1 |
ENST00000265970.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013148.hg.1 |
ENST00000265970.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013148.hg.1 |
ENST00000265970.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013148.hg.1 |
ENST00000265970.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013148.hg.1 |
ENST00000265970.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013148.hg.1 |
ENST00000265970.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013148.hg.1 |
ENST00000265970.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013148.hg.1 |
ENST00000503729.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013148.hg.1 |
ENST00000503729.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013148.hg.1 |
ENST00000503729.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013148.hg.1 |
ENST00000532035.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013148.hg.1 |
ENST00000532035.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013148.hg.1 |
ENST00000532035.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013148.hg.1 |
ENST00000533645.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013148.hg.1 |
ENST00000533645.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013148.hg.1 |
ENST00000533645.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013148.hg.1 |
ENST00000533645.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013148.hg.1 |
ENST00000533645.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013148.hg.1 |
ENST00000533645.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013148.hg.1 |
ENST00000533645.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013148.hg.1 |
ENST00000533645.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013148.hg.1 |
ENST00000533645.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013148.hg.1 |
NM_002645.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013148.hg.1 |
NM_002645.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013148.hg.1 |
NM_002645.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013148.hg.1 |
NM_002645.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013148.hg.1 |
NM_002645.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013148.hg.1 |
NM_002645.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013148.hg.1 |
NM_002645.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013148.hg.1 |
NM_002645.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013148.hg.1 |
NM_002645.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013148.hg.1 |
NM_002645.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013148.hg.1 |
NM_002645.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013148.hg.1 |
NM_002645.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013148.hg.1 |
NM_002645.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013148.hg.1 |
NM_002645.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013148.hg.1 |
NM_002645.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013148.hg.1 |
NM_002645.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013148.hg.1 |
NM_002645.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013148.hg.1 |
NM_002645.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013148.hg.1 |
NM_002645.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013148.hg.1 |
NM_002645.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013148.hg.1 |
NM_002645.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013148.hg.1 |
NM_002645.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013148.hg.1 |
NM_002645.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013148.hg.1 |
NM_002645.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013148.hg.1 |
NM_002645.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013148.hg.1 |
NM_002645.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013148.hg.1 |
NM_002645.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013148.hg.1 |
NM_002645.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013148.hg.1 |
NM_002645.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013148.hg.1 |
NM_002645.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013148.hg.1 |
NM_002645.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013148.hg.1 |
NM_002645.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013148.hg.1 |
NM_002645.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013148.hg.1 |
NM_002645.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013148.hg.1 |
NM_002645.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013148.hg.1 |
NM_002645.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100203742.hg.1 |
ENST00000265970.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100203742.hg.1 |
ENST00000532035.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100203742.hg.1 |
ENST00000533645.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100203742.hg.1 |
NM_002645.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100203742.hg.1 |
ENST00000265970.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100203742.hg.1 |
ENST00000532035.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100203742.hg.1 |
ENST00000533645.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100203742.hg.1 |
NM_002645.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100203742.hg.1 |
TC1100013148.hg.1 |
Clariom D TC |