Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008132.hg.1 |
ENST00000371477.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008132.hg.1 |
ENST00000371477.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008132.hg.1 |
ENST00000371477.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008132.hg.1 |
ENST00000371477.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008132.hg.1 |
ENST00000371477.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008132.hg.1 |
ENST00000371477.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008132.hg.1 |
ENST00000371477.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008132.hg.1 |
ENST00000371477.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008132.hg.1 |
ENST00000371477.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008132.hg.1 |
ENST00000371477.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008132.hg.1 |
ENST00000371477.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008132.hg.1 |
ENST00000371477.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008132.hg.1 |
ENST00000371477.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008132.hg.1 |
ENST00000371477.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008132.hg.1 |
ENST00000371477.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008132.hg.1 |
ENST00000371477.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008132.hg.1 |
ENST00000371477.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008132.hg.1 |
ENST00000371477.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008132.hg.1 |
ENST00000371477.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008132.hg.1 |
ENST00000371477.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008132.hg.1 |
NM_001253.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008132.hg.1 |
NM_001253.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008132.hg.1 |
NM_001253.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008132.hg.1 |
NM_001253.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008132.hg.1 |
NM_001253.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008132.hg.1 |
NM_001253.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008132.hg.1 |
NM_001253.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008132.hg.1 |
NM_001253.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008132.hg.1 |
NM_001253.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008132.hg.1 |
NM_001253.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008132.hg.1 |
NM_001253.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008132.hg.1 |
NM_001253.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008132.hg.1 |
NM_001253.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008132.hg.1 |
NM_001253.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008132.hg.1 |
NM_001253.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008132.hg.1 |
NM_001253.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008132.hg.1 |
NM_001253.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008132.hg.1 |
NM_001253.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008132.hg.1 |
NM_001253.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008132.hg.1 |
NM_001253.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06007538.hg.1 |
TC06000617.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06007538.hg.1 |
ENST00000371477.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06007538.hg.1 |
NM_001253.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600156971.hg.1 |
ENST00000371477.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600156971.hg.1 |
NM_001253.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600156971.hg.1 |
ENST00000371477.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600156971.hg.1 |
NM_001253.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600156971.hg.1 |
TC0600008132.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600008132.hg.1 |
ENST00000371477.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008132.hg.1 |
ENST00000371477.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008132.hg.1 |
ENST00000371477.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008132.hg.1 |
ENST00000371477.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008132.hg.1 |
ENST00000371477.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008132.hg.1 |
ENST00000371477.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008132.hg.1 |
ENST00000371477.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008132.hg.1 |
ENST00000371477.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008132.hg.1 |
ENST00000371477.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008132.hg.1 |
ENST00000371477.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008132.hg.1 |
ENST00000371477.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008132.hg.1 |
ENST00000371477.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008132.hg.1 |
ENST00000371477.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008132.hg.1 |
ENST00000371477.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008132.hg.1 |
ENST00000371477.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008132.hg.1 |
ENST00000371477.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008132.hg.1 |
ENST00000371477.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008132.hg.1 |
ENST00000371477.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008132.hg.1 |
ENST00000371477.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008132.hg.1 |
ENST00000371477.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008132.hg.1 |
NM_001253.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008132.hg.1 |
NM_001253.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008132.hg.1 |
NM_001253.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008132.hg.1 |
NM_001253.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008132.hg.1 |
NM_001253.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008132.hg.1 |
NM_001253.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008132.hg.1 |
NM_001253.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008132.hg.1 |
NM_001253.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008132.hg.1 |
NM_001253.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008132.hg.1 |
NM_001253.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008132.hg.1 |
NM_001253.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008132.hg.1 |
NM_001253.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008132.hg.1 |
NM_001253.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008132.hg.1 |
NM_001253.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008132.hg.1 |
NM_001253.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008132.hg.1 |
NM_001253.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008132.hg.1 |
NM_001253.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008132.hg.1 |
NM_001253.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008132.hg.1 |
NM_001253.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008132.hg.1 |
NM_001253.3 |
Refseq |