Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008141.hg.1 |
ENST00000290858.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008141.hg.1 |
ENST00000290858.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008141.hg.1 |
ENST00000290858.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008141.hg.1 |
ENST00000290858.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008141.hg.1 |
ENST00000290858.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008141.hg.1 |
ENST00000290858.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008141.hg.1 |
ENST00000290858.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008141.hg.1 |
ENST00000290858.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008141.hg.1 |
ENST00000290858.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008141.hg.1 |
ENST00000290858.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008141.hg.1 |
ENST00000290858.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008141.hg.1 |
ENST00000290858.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008141.hg.1 |
ENST00000290858.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008141.hg.1 |
ENST00000290858.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008141.hg.1 |
ENST00000290858.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008141.hg.1 |
ENST00000290858.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008141.hg.1 |
ENST00000290858.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008141.hg.1 |
ENST00000412916.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008141.hg.1 |
ENST00000412916.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008141.hg.1 |
ENST00000412916.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008141.hg.1 |
ENST00000412916.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008141.hg.1 |
ENST00000412916.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008141.hg.1 |
ENST00000412916.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008141.hg.1 |
ENST00000412916.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008141.hg.1 |
ENST00000412916.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008141.hg.1 |
ENST00000412916.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008141.hg.1 |
ENST00000412916.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008141.hg.1 |
ENST00000412916.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008141.hg.1 |
ENST00000412916.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008141.hg.1 |
ENST00000412916.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008141.hg.1 |
ENST00000561924.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008141.hg.1 |
ENST00000561924.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008141.hg.1 |
ENST00000561924.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008141.hg.1 |
ENST00000561924.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008141.hg.1 |
ENST00000561924.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008141.hg.1 |
ENST00000561924.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008141.hg.1 |
ENST00000561924.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008141.hg.1 |
ENST00000561924.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008141.hg.1 |
ENST00000563939.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008141.hg.1 |
ENST00000563939.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008141.hg.1 |
ENST00000563939.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008141.hg.1 |
ENST00000563939.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008141.hg.1 |
ENST00000564034.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008141.hg.1 |
ENST00000564034.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008141.hg.1 |
ENST00000564034.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008141.hg.1 |
ENST00000564034.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008141.hg.1 |
ENST00000564034.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008141.hg.1 |
ENST00000564034.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008141.hg.1 |
ENST00000565389.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008141.hg.1 |
ENST00000565389.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008141.hg.1 |
ENST00000565389.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008141.hg.1 |
ENST00000565389.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008141.hg.1 |
ENST00000565389.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008141.hg.1 |
ENST00000565389.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008141.hg.1 |
ENST00000565389.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008141.hg.1 |
ENST00000566281.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008141.hg.1 |
ENST00000566281.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008141.hg.1 |
ENST00000566281.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008141.hg.1 |
ENST00000566281.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008141.hg.1 |
ENST00000566281.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008141.hg.1 |
ENST00000567947.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008141.hg.1 |
ENST00000567947.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008141.hg.1 |
ENST00000567947.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008141.hg.1 |
ENST00000567947.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008141.hg.1 |
ENST00000568858.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008141.hg.1 |
ENST00000568858.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008141.hg.1 |
ENST00000568858.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008141.hg.1 |
ENST00000568858.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008141.hg.1 |
ENST00000568858.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008141.hg.1 |
NM_001755.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008141.hg.1 |
NM_001755.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008141.hg.1 |
NM_001755.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008141.hg.1 |
NM_001755.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008141.hg.1 |
NM_001755.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008141.hg.1 |
NM_001755.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008141.hg.1 |
NM_001755.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008141.hg.1 |
NM_001755.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008141.hg.1 |
NM_001755.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008141.hg.1 |
NM_001755.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008141.hg.1 |
NM_001755.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008141.hg.1 |
NM_001755.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008141.hg.1 |
NM_001755.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008141.hg.1 |
NM_001755.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008141.hg.1 |
NM_001755.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008141.hg.1 |
NM_001755.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008141.hg.1 |
NM_001755.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008141.hg.1 |
NM_022845.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008141.hg.1 |
NM_022845.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008141.hg.1 |
NM_022845.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008141.hg.1 |
NM_022845.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008141.hg.1 |
NM_022845.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008141.hg.1 |
NM_022845.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008141.hg.1 |
NM_022845.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008141.hg.1 |
NM_022845.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008141.hg.1 |
NM_022845.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008141.hg.1 |
NM_022845.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008141.hg.1 |
NM_022845.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008141.hg.1 |
NM_022845.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008141.hg.1 |
NM_022845.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008141.hg.1 |
NM_022845.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008141.hg.1 |
NM_022845.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008141.hg.1 |
NM_022845.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16007377.hg.1 |
ENST00000290858.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16007377.hg.1 |
ENST00000412916.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16007377.hg.1 |
NM_001755.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16007377.hg.1 |
NM_022845.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16007377.hg.1 |
TC16000528.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1600008141.hg.1 |
ENST00000290858.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008141.hg.1 |
ENST00000290858.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008141.hg.1 |
ENST00000290858.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008141.hg.1 |
ENST00000290858.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008141.hg.1 |
ENST00000290858.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008141.hg.1 |
ENST00000290858.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008141.hg.1 |
ENST00000290858.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008141.hg.1 |
ENST00000290858.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008141.hg.1 |
ENST00000290858.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008141.hg.1 |
ENST00000290858.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008141.hg.1 |
ENST00000290858.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008141.hg.1 |
ENST00000290858.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008141.hg.1 |
ENST00000290858.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008141.hg.1 |
ENST00000290858.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008141.hg.1 |
ENST00000290858.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008141.hg.1 |
ENST00000290858.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008141.hg.1 |
ENST00000290858.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008141.hg.1 |
ENST00000412916.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008141.hg.1 |
ENST00000412916.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008141.hg.1 |
ENST00000412916.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008141.hg.1 |
ENST00000412916.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008141.hg.1 |
ENST00000412916.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008141.hg.1 |
ENST00000412916.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008141.hg.1 |
ENST00000412916.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008141.hg.1 |
ENST00000412916.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008141.hg.1 |
ENST00000412916.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008141.hg.1 |
ENST00000412916.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008141.hg.1 |
ENST00000412916.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008141.hg.1 |
ENST00000412916.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008141.hg.1 |
ENST00000412916.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008141.hg.1 |
ENST00000561924.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008141.hg.1 |
ENST00000561924.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008141.hg.1 |
ENST00000561924.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008141.hg.1 |
ENST00000561924.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008141.hg.1 |
ENST00000561924.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008141.hg.1 |
ENST00000561924.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008141.hg.1 |
ENST00000561924.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008141.hg.1 |
ENST00000561924.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008141.hg.1 |
ENST00000563939.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008141.hg.1 |
ENST00000563939.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008141.hg.1 |
ENST00000563939.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008141.hg.1 |
ENST00000563939.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008141.hg.1 |
ENST00000564034.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008141.hg.1 |
ENST00000564034.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008141.hg.1 |
ENST00000564034.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008141.hg.1 |
ENST00000564034.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008141.hg.1 |
ENST00000564034.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008141.hg.1 |
ENST00000564034.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008141.hg.1 |
ENST00000565389.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008141.hg.1 |
ENST00000565389.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008141.hg.1 |
ENST00000565389.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008141.hg.1 |
ENST00000565389.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008141.hg.1 |
ENST00000565389.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008141.hg.1 |
ENST00000565389.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008141.hg.1 |
ENST00000565389.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008141.hg.1 |
ENST00000566281.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008141.hg.1 |
ENST00000566281.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008141.hg.1 |
ENST00000566281.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008141.hg.1 |
ENST00000566281.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008141.hg.1 |
ENST00000566281.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008141.hg.1 |
ENST00000567947.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008141.hg.1 |
ENST00000567947.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008141.hg.1 |
ENST00000567947.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008141.hg.1 |
ENST00000567947.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008141.hg.1 |
ENST00000568858.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008141.hg.1 |
ENST00000568858.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008141.hg.1 |
ENST00000568858.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008141.hg.1 |
ENST00000568858.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008141.hg.1 |
ENST00000568858.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008141.hg.1 |
NM_001755.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008141.hg.1 |
NM_001755.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008141.hg.1 |
NM_001755.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008141.hg.1 |
NM_001755.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008141.hg.1 |
NM_001755.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008141.hg.1 |
NM_001755.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008141.hg.1 |
NM_001755.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008141.hg.1 |
NM_001755.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008141.hg.1 |
NM_001755.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008141.hg.1 |
NM_001755.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008141.hg.1 |
NM_001755.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008141.hg.1 |
NM_001755.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008141.hg.1 |
NM_001755.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008141.hg.1 |
NM_001755.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008141.hg.1 |
NM_001755.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008141.hg.1 |
NM_001755.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008141.hg.1 |
NM_001755.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008141.hg.1 |
NM_022845.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008141.hg.1 |
NM_022845.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008141.hg.1 |
NM_022845.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008141.hg.1 |
NM_022845.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008141.hg.1 |
NM_022845.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008141.hg.1 |
NM_022845.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008141.hg.1 |
NM_022845.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008141.hg.1 |
NM_022845.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008141.hg.1 |
NM_022845.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008141.hg.1 |
NM_022845.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008141.hg.1 |
NM_022845.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008141.hg.1 |
NM_022845.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008141.hg.1 |
NM_022845.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008141.hg.1 |
NM_022845.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008141.hg.1 |
NM_022845.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008141.hg.1 |
NM_022845.2 |
Refseq |