Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OLIG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138983.2 | NP_620450.2 | 1 | 155 | 70 | |
GenBank mRNA | AB209791.1 | - | 1 | 146 | 70 | |
AL834450.1 | - | 1 | 148 | 70 | ||
BC033290.1 | - | 1 | 162 | 4940343 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |