Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007244.hg.1 |
ENST00000334979.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007244.hg.1 |
ENST00000334979.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007244.hg.1 |
ENST00000334979.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007244.hg.1 |
ENST00000334979.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007244.hg.1 |
ENST00000334979.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007244.hg.1 |
ENST00000334979.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007244.hg.1 |
ENST00000334979.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007244.hg.1 |
ENST00000334979.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007244.hg.1 |
ENST00000334979.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007244.hg.1 |
ENST00000377961.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007244.hg.1 |
ENST00000377961.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007244.hg.1 |
ENST00000377961.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007244.hg.1 |
ENST00000377961.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007244.hg.1 |
ENST00000377961.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007244.hg.1 |
ENST00000377961.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007244.hg.1 |
ENST00000377961.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007244.hg.1 |
ENST00000377961.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007244.hg.1 |
ENST00000377961.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007244.hg.1 |
ENST00000377961.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007244.hg.1 |
ENST00000377961.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007244.hg.1 |
ENST00000377961.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007244.hg.1 |
ENST00000377961.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007244.hg.1 |
ENST00000612225.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007244.hg.1 |
ENST00000612225.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007244.hg.1 |
ENST00000612225.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007244.hg.1 |
ENST00000612225.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007244.hg.1 |
ENST00000612225.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007244.hg.1 |
ENST00000612225.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007244.hg.1 |
ENST00000612225.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007244.hg.1 |
ENST00000612225.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007244.hg.1 |
ENST00000612225.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007244.hg.1 |
ENST00000612225.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007244.hg.1 |
ENST00000612225.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007244.hg.1 |
ENST00000612225.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007244.hg.1 |
ENST00000612225.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007244.hg.1 |
NM_006998.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007244.hg.1 |
NM_006998.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007244.hg.1 |
NM_006998.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007244.hg.1 |
NM_006998.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007244.hg.1 |
NM_006998.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007244.hg.1 |
NM_006998.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007244.hg.1 |
NM_006998.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007244.hg.1 |
NM_006998.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007244.hg.1 |
NM_006998.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007244.hg.1 |
NM_006998.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007244.hg.1 |
NM_006998.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007244.hg.1 |
NM_006998.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007244.hg.1 |
NM_006998.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007244.hg.1 |
NM_006998.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06001597.hg.1 |
TC06000157.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06001597.hg.1 |
ENST00000334979.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06001597.hg.1 |
ENST00000377961.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06001597.hg.1 |
NM_006998.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600149161.hg.1 |
ENST00000334979.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600149161.hg.1 |
ENST00000377961.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600149161.hg.1 |
ENST00000612225.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600149161.hg.1 |
NM_006998.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600149161.hg.1 |
ENST00000334979.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600149161.hg.1 |
ENST00000377961.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600149161.hg.1 |
ENST00000612225.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600149161.hg.1 |
NM_006998.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600149161.hg.1 |
TC0600007244.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600007244.hg.1 |
ENST00000334979.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007244.hg.1 |
ENST00000334979.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007244.hg.1 |
ENST00000334979.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007244.hg.1 |
ENST00000334979.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007244.hg.1 |
ENST00000334979.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007244.hg.1 |
ENST00000334979.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007244.hg.1 |
ENST00000334979.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007244.hg.1 |
ENST00000334979.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007244.hg.1 |
ENST00000334979.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007244.hg.1 |
ENST00000377961.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007244.hg.1 |
ENST00000377961.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007244.hg.1 |
ENST00000377961.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007244.hg.1 |
ENST00000377961.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007244.hg.1 |
ENST00000377961.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007244.hg.1 |
ENST00000377961.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007244.hg.1 |
ENST00000377961.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007244.hg.1 |
ENST00000377961.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007244.hg.1 |
ENST00000377961.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007244.hg.1 |
ENST00000377961.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007244.hg.1 |
ENST00000377961.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007244.hg.1 |
ENST00000377961.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007244.hg.1 |
ENST00000377961.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007244.hg.1 |
ENST00000612225.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007244.hg.1 |
ENST00000612225.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007244.hg.1 |
ENST00000612225.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007244.hg.1 |
ENST00000612225.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007244.hg.1 |
ENST00000612225.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007244.hg.1 |
ENST00000612225.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007244.hg.1 |
ENST00000612225.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007244.hg.1 |
ENST00000612225.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007244.hg.1 |
ENST00000612225.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007244.hg.1 |
ENST00000612225.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007244.hg.1 |
ENST00000612225.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007244.hg.1 |
ENST00000612225.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007244.hg.1 |
ENST00000612225.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007244.hg.1 |
NM_006998.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007244.hg.1 |
NM_006998.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007244.hg.1 |
NM_006998.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007244.hg.1 |
NM_006998.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007244.hg.1 |
NM_006998.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007244.hg.1 |
NM_006998.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007244.hg.1 |
NM_006998.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007244.hg.1 |
NM_006998.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007244.hg.1 |
NM_006998.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007244.hg.1 |
NM_006998.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007244.hg.1 |
NM_006998.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007244.hg.1 |
NM_006998.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007244.hg.1 |
NM_006998.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007244.hg.1 |
NM_006998.3 |
Refseq |