Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X015733.hg.1 |
TC0X001165.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X015733.hg.1 |
ENST00000373361.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X015733.hg.1 |
ENST00000373367.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X015733.hg.1 |
ENST00000482827.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X015733.hg.1 |
ENST00000541184.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X015733.hg.1 |
NM_001146256.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X015733.hg.1 |
NM_001146257.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X015733.hg.1 |
NM_144969.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X015730.hg.1 |
TC0X001165.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X015730.hg.1 |
ENST00000373361.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X015730.hg.1 |
ENST00000373367.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X015730.hg.1 |
ENST00000541184.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X015730.hg.1 |
NM_001146256.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X015730.hg.1 |
NM_001146257.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X015730.hg.1 |
NM_144969.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X015734.hg.1 |
TC0X001165.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X015734.hg.1 |
ENST00000373361.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X015734.hg.1 |
ENST00000373367.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X015734.hg.1 |
ENST00000541184.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X015734.hg.1 |
NM_001146256.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X015734.hg.1 |
NM_001146257.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X015734.hg.1 |
NM_144969.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X015737.hg.1 |
TC0X001165.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X015737.hg.1 |
ENST00000541184.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X015737.hg.1 |
NM_001146256.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X015737.hg.1 |
NM_001146257.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X015737.hg.1 |
NM_144969.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X015736.hg.1 |
TC0X001165.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X015736.hg.1 |
ENST00000373367.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X015736.hg.1 |
ENST00000541184.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X015736.hg.1 |
NM_001146256.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X015736.hg.1 |
NM_001146257.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X015736.hg.1 |
NM_144969.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010107.hg.1 |
ENST00000373367.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010107.hg.1 |
ENST00000373367.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010107.hg.1 |
ENST00000373367.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010107.hg.1 |
ENST00000373367.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010107.hg.1 |
ENST00000373367.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010107.hg.1 |
ENST00000373367.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010107.hg.1 |
ENST00000373367.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010107.hg.1 |
ENST00000373367.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010107.hg.1 |
ENST00000373367.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010107.hg.1 |
ENST00000373367.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010107.hg.1 |
ENST00000373367.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010107.hg.1 |
ENST00000373367.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010107.hg.1 |
ENST00000373367.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010107.hg.1 |
ENST00000373367.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010107.hg.1 |
ENST00000373367.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010107.hg.1 |
ENST00000373367.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010107.hg.1 |
ENST00000482827.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010107.hg.1 |
ENST00000482827.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010107.hg.1 |
ENST00000541184.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010107.hg.1 |
ENST00000541184.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010107.hg.1 |
ENST00000541184.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010107.hg.1 |
ENST00000541184.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010107.hg.1 |
ENST00000541184.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010107.hg.1 |
ENST00000541184.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010107.hg.1 |
ENST00000541184.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010107.hg.1 |
ENST00000541184.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010107.hg.1 |
ENST00000541184.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010107.hg.1 |
ENST00000541184.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010107.hg.1 |
ENST00000541184.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010107.hg.1 |
ENST00000541184.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010107.hg.1 |
ENST00000541184.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010107.hg.1 |
ENST00000541184.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010107.hg.1 |
ENST00000541184.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010107.hg.1 |
ENST00000541184.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010107.hg.1 |
NM_001146256.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010107.hg.1 |
NM_001146256.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010107.hg.1 |
NM_001146256.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010107.hg.1 |
NM_001146256.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010107.hg.1 |
NM_001146256.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010107.hg.1 |
NM_001146256.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010107.hg.1 |
NM_001146256.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010107.hg.1 |
NM_001146256.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010107.hg.1 |
NM_001146256.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010107.hg.1 |
NM_001146256.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010107.hg.1 |
NM_001146256.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010107.hg.1 |
NM_001146256.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010107.hg.1 |
NM_001146256.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010107.hg.1 |
NM_001146256.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010107.hg.1 |
NM_001146256.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010107.hg.1 |
NM_001146256.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010107.hg.1 |
NM_001146257.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010107.hg.1 |
NM_001146257.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010107.hg.1 |
NM_001146257.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010107.hg.1 |
NM_001146257.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010107.hg.1 |
NM_001146257.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010107.hg.1 |
NM_001146257.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010107.hg.1 |
NM_144969.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010107.hg.1 |
NM_144969.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010107.hg.1 |
NM_144969.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010107.hg.1 |
NM_144969.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010107.hg.1 |
NM_144969.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010107.hg.1 |
NM_144969.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010107.hg.1 |
NM_144969.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010107.hg.1 |
NM_144969.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010107.hg.1 |
NM_144969.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010107.hg.1 |
NM_144969.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010107.hg.1 |
NM_144969.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010107.hg.1 |
NM_144969.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010107.hg.1 |
NM_144969.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010107.hg.1 |
NM_144969.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010107.hg.1 |
NM_144969.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010107.hg.1 |
NM_144969.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010107.hg.1 |
NM_144969.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00168990.hg.1 |
ENST00000373367.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00168990.hg.1 |
ENST00000482827.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00168990.hg.1 |
ENST00000541184.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00168990.hg.1 |
NM_001146256.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00168990.hg.1 |
NM_001146257.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00168990.hg.1 |
NM_144969.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00168990.hg.1 |
ENST00000373367.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00168990.hg.1 |
ENST00000482827.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00168990.hg.1 |
ENST00000541184.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00168990.hg.1 |
NM_001146256.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00168990.hg.1 |
NM_001146257.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00168990.hg.1 |
NM_144969.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00168990.hg.1 |
TC0X00010107.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00168994.hg.1 |
ENST00000541184.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00168994.hg.1 |
NM_001146256.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00168994.hg.1 |
NM_001146257.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00168994.hg.1 |
NM_144969.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00168994.hg.1 |
ENST00000541184.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00168994.hg.1 |
NM_001146256.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00168994.hg.1 |
NM_001146257.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00168994.hg.1 |
NM_144969.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00168994.hg.1 |
TC0X00010107.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0X00010107.hg.1 |
ENST00000373367.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010107.hg.1 |
ENST00000373367.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010107.hg.1 |
ENST00000373367.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010107.hg.1 |
ENST00000373367.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010107.hg.1 |
ENST00000373367.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010107.hg.1 |
ENST00000373367.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010107.hg.1 |
ENST00000373367.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010107.hg.1 |
ENST00000373367.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010107.hg.1 |
ENST00000373367.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010107.hg.1 |
ENST00000373367.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010107.hg.1 |
ENST00000373367.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010107.hg.1 |
ENST00000373367.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010107.hg.1 |
ENST00000373367.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010107.hg.1 |
ENST00000373367.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010107.hg.1 |
ENST00000373367.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010107.hg.1 |
ENST00000373367.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010107.hg.1 |
ENST00000482827.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010107.hg.1 |
ENST00000482827.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010107.hg.1 |
ENST00000541184.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010107.hg.1 |
ENST00000541184.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010107.hg.1 |
ENST00000541184.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010107.hg.1 |
ENST00000541184.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010107.hg.1 |
ENST00000541184.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010107.hg.1 |
ENST00000541184.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010107.hg.1 |
ENST00000541184.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010107.hg.1 |
ENST00000541184.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010107.hg.1 |
ENST00000541184.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010107.hg.1 |
ENST00000541184.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010107.hg.1 |
ENST00000541184.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010107.hg.1 |
ENST00000541184.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010107.hg.1 |
ENST00000541184.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010107.hg.1 |
ENST00000541184.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010107.hg.1 |
ENST00000541184.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010107.hg.1 |
ENST00000541184.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010107.hg.1 |
NM_001146256.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010107.hg.1 |
NM_001146256.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010107.hg.1 |
NM_001146256.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010107.hg.1 |
NM_001146256.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010107.hg.1 |
NM_001146256.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010107.hg.1 |
NM_001146256.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010107.hg.1 |
NM_001146256.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010107.hg.1 |
NM_001146256.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010107.hg.1 |
NM_001146256.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010107.hg.1 |
NM_001146256.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010107.hg.1 |
NM_001146256.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010107.hg.1 |
NM_001146256.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010107.hg.1 |
NM_001146256.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010107.hg.1 |
NM_001146256.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010107.hg.1 |
NM_001146256.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010107.hg.1 |
NM_001146256.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010107.hg.1 |
NM_001146257.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010107.hg.1 |
NM_001146257.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010107.hg.1 |
NM_001146257.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010107.hg.1 |
NM_001146257.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010107.hg.1 |
NM_001146257.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010107.hg.1 |
NM_001146257.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010107.hg.1 |
NM_144969.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010107.hg.1 |
NM_144969.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010107.hg.1 |
NM_144969.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010107.hg.1 |
NM_144969.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010107.hg.1 |
NM_144969.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010107.hg.1 |
NM_144969.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010107.hg.1 |
NM_144969.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010107.hg.1 |
NM_144969.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010107.hg.1 |
NM_144969.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010107.hg.1 |
NM_144969.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010107.hg.1 |
NM_144969.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010107.hg.1 |
NM_144969.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010107.hg.1 |
NM_144969.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010107.hg.1 |
NM_144969.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010107.hg.1 |
NM_144969.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010107.hg.1 |
NM_144969.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010107.hg.1 |
NM_144969.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00168987.hg.1 |
ENST00000373367.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00168987.hg.1 |
ENST00000541184.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00168987.hg.1 |
NM_001146256.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00168987.hg.1 |
NM_001146257.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00168987.hg.1 |
NM_144969.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00168987.hg.1 |
ENST00000373367.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00168987.hg.1 |
ENST00000541184.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00168987.hg.1 |
NM_001146256.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00168987.hg.1 |
NM_001146257.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00168987.hg.1 |
NM_144969.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00168987.hg.1 |
TC0X00010107.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00168993.hg.1 |
ENST00000373367.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00168993.hg.1 |
ENST00000541184.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00168993.hg.1 |
NM_001146256.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00168993.hg.1 |
NM_001146257.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00168993.hg.1 |
NM_144969.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00168993.hg.1 |
ENST00000373367.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00168993.hg.1 |
ENST00000541184.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00168993.hg.1 |
NM_001146256.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00168993.hg.1 |
NM_001146257.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00168993.hg.1 |
NM_144969.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00168993.hg.1 |
TC0X00010107.hg.1 |
Clariom D TC |