Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009691.hg.1 |
ENST00000256876.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009691.hg.1 |
ENST00000256876.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009691.hg.1 |
ENST00000256876.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009691.hg.1 |
ENST00000256876.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009691.hg.1 |
ENST00000256876.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009691.hg.1 |
ENST00000256876.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009691.hg.1 |
ENST00000256876.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009691.hg.1 |
ENST00000256876.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009691.hg.1 |
ENST00000256876.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009691.hg.1 |
ENST00000256876.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009691.hg.1 |
ENST00000379954.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009691.hg.1 |
ENST00000379954.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009691.hg.1 |
ENST00000379954.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009691.hg.1 |
ENST00000379954.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009691.hg.1 |
ENST00000379954.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009691.hg.1 |
ENST00000379954.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009691.hg.1 |
ENST00000379954.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009691.hg.1 |
ENST00000379954.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009691.hg.1 |
ENST00000379954.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009691.hg.1 |
ENST00000379954.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009691.hg.1 |
ENST00000379959.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009691.hg.1 |
ENST00000379959.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009691.hg.1 |
ENST00000379959.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009691.hg.1 |
ENST00000379959.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009691.hg.1 |
ENST00000379959.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009691.hg.1 |
ENST00000379959.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009691.hg.1 |
ENST00000379959.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009691.hg.1 |
ENST00000379959.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009691.hg.1 |
ENST00000379959.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009691.hg.1 |
ENST00000379959.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009691.hg.1 |
ENST00000379959.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009691.hg.1 |
ENST00000379959.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009691.hg.1 |
ENST00000379959.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009691.hg.1 |
ENST00000447847.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009691.hg.1 |
ENST00000447847.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009691.hg.1 |
ENST00000447847.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009691.hg.1 |
NM_000417.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009691.hg.1 |
NM_000417.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009691.hg.1 |
NM_000417.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009691.hg.1 |
NM_000417.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009691.hg.1 |
NM_000417.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009691.hg.1 |
NM_000417.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009691.hg.1 |
NM_000417.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009691.hg.1 |
NM_000417.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009691.hg.1 |
NM_000417.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009691.hg.1 |
NM_000417.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009691.hg.1 |
NM_000417.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009691.hg.1 |
NM_000417.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009691.hg.1 |
NM_000417.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009691.hg.1 |
NM_000417.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009691.hg.1 |
NM_001308242.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009691.hg.1 |
NM_001308242.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009691.hg.1 |
NM_001308242.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009691.hg.1 |
NM_001308242.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009691.hg.1 |
NM_001308242.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009691.hg.1 |
NM_001308242.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009691.hg.1 |
NM_001308242.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009691.hg.1 |
NM_001308242.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009691.hg.1 |
NM_001308242.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009691.hg.1 |
NM_001308242.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009691.hg.1 |
NM_001308242.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009691.hg.1 |
NM_001308242.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009691.hg.1 |
NM_001308243.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009691.hg.1 |
NM_001308243.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009691.hg.1 |
NM_001308243.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009691.hg.1 |
NM_001308243.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009691.hg.1 |
NM_001308243.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009691.hg.1 |
NM_001308243.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009691.hg.1 |
NM_001308243.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009691.hg.1 |
NM_001308243.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009691.hg.1 |
NM_001308243.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009691.hg.1 |
NM_001308243.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000009691.hg.1 |
NM_001308243.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10012582.hg.1 |
ENST00000256876.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10012582.hg.1 |
ENST00000379954.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10012582.hg.1 |
ENST00000379959.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10012582.hg.1 |
NM_000417.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10012582.hg.1 |
TC10001007.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR10012583.hg.1 |
ENST00000379959.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10012583.hg.1 |
NM_000417.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10012583.hg.1 |
TC10001007.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1000166256.hg.1 |
ENST00000379959.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000166256.hg.1 |
NM_000417.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000166256.hg.1 |
ENST00000379959.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000166256.hg.1 |
NM_000417.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000166256.hg.1 |
TC1000009691.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1000009691.hg.1 |
ENST00000256876.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009691.hg.1 |
ENST00000256876.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009691.hg.1 |
ENST00000256876.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009691.hg.1 |
ENST00000256876.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009691.hg.1 |
ENST00000256876.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009691.hg.1 |
ENST00000256876.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009691.hg.1 |
ENST00000256876.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009691.hg.1 |
ENST00000256876.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009691.hg.1 |
ENST00000256876.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009691.hg.1 |
ENST00000256876.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009691.hg.1 |
ENST00000379954.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009691.hg.1 |
ENST00000379954.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009691.hg.1 |
ENST00000379954.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009691.hg.1 |
ENST00000379954.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009691.hg.1 |
ENST00000379954.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009691.hg.1 |
ENST00000379954.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009691.hg.1 |
ENST00000379954.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009691.hg.1 |
ENST00000379954.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009691.hg.1 |
ENST00000379954.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009691.hg.1 |
ENST00000379954.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009691.hg.1 |
ENST00000379959.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009691.hg.1 |
ENST00000379959.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009691.hg.1 |
ENST00000379959.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009691.hg.1 |
ENST00000379959.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009691.hg.1 |
ENST00000379959.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009691.hg.1 |
ENST00000379959.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009691.hg.1 |
ENST00000379959.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009691.hg.1 |
ENST00000379959.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009691.hg.1 |
ENST00000379959.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009691.hg.1 |
ENST00000379959.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009691.hg.1 |
ENST00000379959.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009691.hg.1 |
ENST00000379959.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009691.hg.1 |
ENST00000379959.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009691.hg.1 |
ENST00000447847.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009691.hg.1 |
ENST00000447847.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009691.hg.1 |
ENST00000447847.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000009691.hg.1 |
NM_000417.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009691.hg.1 |
NM_000417.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009691.hg.1 |
NM_000417.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009691.hg.1 |
NM_000417.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009691.hg.1 |
NM_000417.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009691.hg.1 |
NM_000417.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009691.hg.1 |
NM_000417.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009691.hg.1 |
NM_000417.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009691.hg.1 |
NM_000417.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009691.hg.1 |
NM_000417.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009691.hg.1 |
NM_000417.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009691.hg.1 |
NM_000417.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009691.hg.1 |
NM_000417.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009691.hg.1 |
NM_000417.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009691.hg.1 |
NM_001308242.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009691.hg.1 |
NM_001308242.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009691.hg.1 |
NM_001308242.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009691.hg.1 |
NM_001308242.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009691.hg.1 |
NM_001308242.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009691.hg.1 |
NM_001308242.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009691.hg.1 |
NM_001308242.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009691.hg.1 |
NM_001308242.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009691.hg.1 |
NM_001308242.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009691.hg.1 |
NM_001308242.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009691.hg.1 |
NM_001308242.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009691.hg.1 |
NM_001308242.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009691.hg.1 |
NM_001308243.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009691.hg.1 |
NM_001308243.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009691.hg.1 |
NM_001308243.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009691.hg.1 |
NM_001308243.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009691.hg.1 |
NM_001308243.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009691.hg.1 |
NM_001308243.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009691.hg.1 |
NM_001308243.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009691.hg.1 |
NM_001308243.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009691.hg.1 |
NM_001308243.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009691.hg.1 |
NM_001308243.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000009691.hg.1 |
NM_001308243.1 |
Refseq |