Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007622.hg.1 |
ENST00000262053.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007622.hg.1 |
ENST00000262053.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007622.hg.1 |
ENST00000262053.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007622.hg.1 |
ENST00000262053.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007622.hg.1 |
ENST00000262053.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007622.hg.1 |
ENST00000262053.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007622.hg.1 |
ENST00000262053.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007622.hg.1 |
ENST00000262053.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007622.hg.1 |
ENST00000262053.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007622.hg.1 |
ENST00000262053.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007622.hg.1 |
ENST00000549612.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007622.hg.1 |
ENST00000549612.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007622.hg.1 |
ENST00000549612.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007622.hg.1 |
ENST00000549612.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007622.hg.1 |
ENST00000551831.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007622.hg.1 |
ENST00000551831.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007622.hg.1 |
ENST00000551831.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007622.hg.1 |
ENST00000551831.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007622.hg.1 |
ENST00000551831.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007622.hg.1 |
ENST00000551831.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007622.hg.1 |
ENST00000551831.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007622.hg.1 |
ENST00000551831.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007622.hg.1 |
ENST00000551831.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007622.hg.1 |
ENST00000552487.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007622.hg.1 |
ENST00000552487.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007622.hg.1 |
ENST00000552487.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007622.hg.1 |
ENST00000552487.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007622.hg.1 |
ENST00000552487.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007622.hg.1 |
ENST00000552487.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007622.hg.1 |
ENST00000552487.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007622.hg.1 |
ENST00000552510.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007622.hg.1 |
ENST00000552510.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007622.hg.1 |
ENST00000552510.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007622.hg.1 |
ENST00000552510.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007622.hg.1 |
ENST00000552510.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007622.hg.1 |
NM_005171.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007622.hg.1 |
NM_005171.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007622.hg.1 |
NM_005171.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007622.hg.1 |
NM_005171.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007622.hg.1 |
NM_005171.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007622.hg.1 |
NM_005171.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007622.hg.1 |
NM_005171.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007622.hg.1 |
NM_005171.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007622.hg.1 |
NM_005171.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007622.hg.1 |
NM_005171.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007622.hg.1 |
NM_005171.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007622.hg.1 |
NM_005171.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007622.hg.1 |
NM_005171.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12004988.hg.1 |
ENST00000262053.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12004988.hg.1 |
NM_005171.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12004988.hg.1 |
TC12000397.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1200154002.hg.1 |
ENST00000262053.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200154002.hg.1 |
ENST00000552487.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200154002.hg.1 |
ENST00000552510.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200154002.hg.1 |
NM_005171.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200154002.hg.1 |
ENST00000262053.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200154002.hg.1 |
ENST00000552487.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200154002.hg.1 |
ENST00000552510.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200154002.hg.1 |
NM_005171.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200154002.hg.1 |
TC1200007622.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1200007622.hg.1 |
ENST00000262053.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007622.hg.1 |
ENST00000262053.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007622.hg.1 |
ENST00000262053.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007622.hg.1 |
ENST00000262053.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007622.hg.1 |
ENST00000262053.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007622.hg.1 |
ENST00000262053.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007622.hg.1 |
ENST00000262053.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007622.hg.1 |
ENST00000262053.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007622.hg.1 |
ENST00000262053.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007622.hg.1 |
ENST00000262053.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007622.hg.1 |
ENST00000549612.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007622.hg.1 |
ENST00000549612.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007622.hg.1 |
ENST00000549612.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007622.hg.1 |
ENST00000549612.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007622.hg.1 |
ENST00000551831.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007622.hg.1 |
ENST00000551831.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007622.hg.1 |
ENST00000551831.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007622.hg.1 |
ENST00000551831.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007622.hg.1 |
ENST00000551831.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007622.hg.1 |
ENST00000551831.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007622.hg.1 |
ENST00000551831.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007622.hg.1 |
ENST00000551831.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007622.hg.1 |
ENST00000551831.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007622.hg.1 |
ENST00000552487.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007622.hg.1 |
ENST00000552487.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007622.hg.1 |
ENST00000552487.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007622.hg.1 |
ENST00000552487.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007622.hg.1 |
ENST00000552487.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007622.hg.1 |
ENST00000552487.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007622.hg.1 |
ENST00000552487.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007622.hg.1 |
ENST00000552510.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007622.hg.1 |
ENST00000552510.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007622.hg.1 |
ENST00000552510.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007622.hg.1 |
ENST00000552510.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007622.hg.1 |
ENST00000552510.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007622.hg.1 |
NM_005171.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007622.hg.1 |
NM_005171.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007622.hg.1 |
NM_005171.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007622.hg.1 |
NM_005171.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007622.hg.1 |
NM_005171.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007622.hg.1 |
NM_005171.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007622.hg.1 |
NM_005171.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007622.hg.1 |
NM_005171.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007622.hg.1 |
NM_005171.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007622.hg.1 |
NM_005171.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007622.hg.1 |
NM_005171.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007622.hg.1 |
NM_005171.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007622.hg.1 |
NM_005171.4 |
Refseq |