Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01027547.hg.1 |
TC01001755.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01027547.hg.1 |
ENST00000261458.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01027547.hg.1 |
ENST00000308852.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01027547.hg.1 |
ENST00000367010.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01027547.hg.1 |
ENST00000391905.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01027547.hg.1 |
ENST00000413764.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01027547.hg.1 |
ENST00000537898.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01027547.hg.1 |
ENST00000545154.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01027547.hg.1 |
ENST00000545781.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01027547.hg.1 |
NM_001122834.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01027547.hg.1 |
NM_001170580.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01027547.hg.1 |
NM_001170587.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01027547.hg.1 |
NM_001170588.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01027547.hg.1 |
NM_018194.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100184565.hg.1 |
ENST00000261458.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100184565.hg.1 |
ENST00000367010.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100184565.hg.1 |
ENST00000413764.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100184565.hg.1 |
ENST00000426968.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100184565.hg.1 |
ENST00000537898.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100184565.hg.1 |
ENST00000545154.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100184565.hg.1 |
NM_001122834.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100184565.hg.1 |
NM_001170580.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100184565.hg.1 |
NM_001170587.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100184565.hg.1 |
NM_001170588.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100184565.hg.1 |
NM_018194.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100184565.hg.1 |
ENST00000261458.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100184565.hg.1 |
ENST00000367010.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100184565.hg.1 |
ENST00000413764.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100184565.hg.1 |
ENST00000426968.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100184565.hg.1 |
ENST00000537898.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100184565.hg.1 |
ENST00000545154.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100184565.hg.1 |
NM_001122834.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100184565.hg.1 |
NM_001170580.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100184565.hg.1 |
NM_001170587.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100184565.hg.1 |
NM_001170588.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100184565.hg.1 |
NM_018194.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100184565.hg.1 |
TC0100011472.hg.1 |
Clariom D TC |