Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000254799.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000254799.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000254799.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000254799.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000254799.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000254799.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000254799.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000254799.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000254799.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000254799.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000254799.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000254799.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000254799.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000254799.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000254799.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000254799.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000254799.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000254799.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000254799.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000254799.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000254799.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000254799.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000254799.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000254799.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000499044.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000499044.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000499044.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000499044.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000499044.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000499044.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000499044.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000499044.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000499044.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000499044.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000499044.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000499044.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000499044.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000499044.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000499044.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000499044.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000499044.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000499044.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000499044.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000499044.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000502323.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000502323.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000502323.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000502323.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000502323.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000502323.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000502323.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000502323.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000502323.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000502323.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000502323.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000502323.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000502323.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000502323.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000502323.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000502323.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000505068.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000505068.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000505068.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000505068.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000505068.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000505068.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000505068.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000505068.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000505068.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000505068.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000505068.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000505068.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000505068.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000505068.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000505068.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000505068.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000505068.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000505068.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000505068.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000505068.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000505068.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000505068.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000505068.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000506453.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000506453.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000506453.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000506453.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000506453.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000506453.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000506453.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000506453.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000506453.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000508091.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000508091.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000508091.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000508091.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000508091.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000514161.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000514161.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000514161.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000514161.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000514161.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000514161.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000514161.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000514161.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000514161.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000514161.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000514161.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000514161.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000514161.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000514161.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000514161.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000514161.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000545193.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000545193.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000545193.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000545193.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000545193.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000545193.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000545193.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000545193.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000545193.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000545193.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000545193.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000545193.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000545193.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000545193.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000545193.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000545193.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000545193.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000545193.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000545193.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000545193.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
ENST00000545193.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
NM_001098477.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
NM_001098477.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
NM_001098477.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
NM_001098477.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
NM_001098477.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
NM_001098477.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
NM_001098477.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
NM_001098477.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
NM_001098477.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
NM_001098477.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
NM_001098477.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
NM_001098477.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
NM_001098477.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
NM_001098477.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
NM_001098477.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
NM_001098477.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
NM_001098477.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
NM_001098477.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
NM_001098477.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
NM_001098477.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
NM_001098477.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
NM_001098477.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
NM_002092.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
NM_002092.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
NM_002092.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
NM_002092.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
NM_002092.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
NM_002092.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
NM_002092.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
NM_002092.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
NM_002092.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
NM_002092.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
NM_002092.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
NM_002092.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
NM_002092.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
NM_002092.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
NM_002092.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
NM_002092.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
NM_002092.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
NM_002092.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
NM_002092.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
NM_002092.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
NM_002092.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
NM_002092.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010965.hg.1 |
NM_002092.3 |
Refseq |