Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011335.hg.1 |
ENST00000510601.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008160.hg.1 |
ENST00000282020.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008160.hg.1 |
ENST00000282020.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008160.hg.1 |
ENST00000282020.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008160.hg.1 |
ENST00000282020.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008160.hg.1 |
ENST00000282020.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008160.hg.1 |
ENST00000282020.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008160.hg.1 |
ENST00000282020.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008160.hg.1 |
ENST00000282020.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008160.hg.1 |
ENST00000282020.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008160.hg.1 |
ENST00000282020.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008160.hg.1 |
ENST00000282020.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008160.hg.1 |
ENST00000282020.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008160.hg.1 |
ENST00000282020.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008160.hg.1 |
ENST00000282020.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008160.hg.1 |
ENST00000282020.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008160.hg.1 |
ENST00000282020.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008160.hg.1 |
ENST00000282020.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008160.hg.1 |
ENST00000282020.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008160.hg.1 |
ENST00000282020.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008160.hg.1 |
ENST00000282020.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008160.hg.1 |
ENST00000282020.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008160.hg.1 |
ENST00000502699.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008160.hg.1 |
ENST00000502699.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008160.hg.1 |
ENST00000502699.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008160.hg.1 |
ENST00000502699.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008160.hg.1 |
ENST00000502699.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008160.hg.1 |
ENST00000505687.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008160.hg.1 |
ENST00000505687.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008160.hg.1 |
ENST00000505687.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008160.hg.1 |
ENST00000505687.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008160.hg.1 |
ENST00000505687.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008160.hg.1 |
ENST00000505687.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008160.hg.1 |
ENST00000505687.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008160.hg.1 |
ENST00000505687.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008160.hg.1 |
ENST00000510992.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008160.hg.1 |
ENST00000510992.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008160.hg.1 |
ENST00000510992.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008160.hg.1 |
ENST00000510992.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008160.hg.1 |
ENST00000510992.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008160.hg.1 |
ENST00000510992.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008160.hg.1 |
ENST00000510992.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008160.hg.1 |
ENST00000510992.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008160.hg.1 |
ENST00000510992.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008160.hg.1 |
ENST00000510992.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008160.hg.1 |
ENST00000510992.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008160.hg.1 |
ENST00000510992.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008160.hg.1 |
ENST00000510992.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008160.hg.1 |
ENST00000510992.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008160.hg.1 |
ENST00000510992.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008160.hg.1 |
ENST00000510992.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008160.hg.1 |
ENST00000512631.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008160.hg.1 |
ENST00000512631.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008160.hg.1 |
ENST00000512631.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008160.hg.1 |
ENST00000512631.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008160.hg.1 |
ENST00000512631.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008160.hg.1 |
ENST00000513976.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008160.hg.1 |
ENST00000513976.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008160.hg.1 |
ENST00000513976.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008160.hg.1 |
ENST00000513976.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008160.hg.1 |
ENST00000513976.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008160.hg.1 |
ENST00000515744.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008160.hg.1 |
ENST00000515744.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008160.hg.1 |
ENST00000515744.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008160.hg.1 |
ENST00000515744.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008160.hg.1 |
ENST00000611049.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008160.hg.1 |
ENST00000611049.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008160.hg.1 |
ENST00000611049.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008160.hg.1 |
ENST00000611049.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008160.hg.1 |
ENST00000611049.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008160.hg.1 |
ENST00000611049.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008160.hg.1 |
ENST00000611049.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008160.hg.1 |
ENST00000611049.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008160.hg.1 |
ENST00000611049.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008160.hg.1 |
ENST00000611049.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008160.hg.1 |
ENST00000611049.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008160.hg.1 |
ENST00000611049.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008160.hg.1 |
ENST00000611049.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008160.hg.1 |
ENST00000611049.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008160.hg.1 |
ENST00000611049.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008160.hg.1 |
ENST00000611049.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008160.hg.1 |
NM_001286838.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008160.hg.1 |
NM_001286838.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008160.hg.1 |
NM_001286838.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008160.hg.1 |
NM_001286838.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008160.hg.1 |
NM_001286838.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008160.hg.1 |
NM_001286838.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008160.hg.1 |
NM_001286838.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008160.hg.1 |
NM_001286838.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008160.hg.1 |
NM_001286838.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008160.hg.1 |
NM_001286838.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008160.hg.1 |
NM_001286838.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008160.hg.1 |
NM_001286838.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008160.hg.1 |
NM_001286838.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008160.hg.1 |
NM_001286838.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008160.hg.1 |
NM_001286838.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008160.hg.1 |
NM_001286838.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008160.hg.1 |
NM_001286838.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008160.hg.1 |
NM_001286838.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008160.hg.1 |
NM_001286838.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008160.hg.1 |
NM_001510.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008160.hg.1 |
NM_001510.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008160.hg.1 |
NM_001510.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008160.hg.1 |
NM_001510.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008160.hg.1 |
NM_001510.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008160.hg.1 |
NM_001510.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008160.hg.1 |
NM_001510.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008160.hg.1 |
NM_001510.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008160.hg.1 |
NM_001510.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008160.hg.1 |
NM_001510.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008160.hg.1 |
NM_001510.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008160.hg.1 |
NM_001510.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008160.hg.1 |
NM_001510.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008160.hg.1 |
NM_001510.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008160.hg.1 |
NM_001510.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008160.hg.1 |
NM_001510.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008160.hg.1 |
NM_001510.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008160.hg.1 |
NM_001510.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008160.hg.1 |
NM_001510.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400008160.hg.1 |
NM_001510.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011335.hg.1 |
ENST00000510601.2 |
Ensembl |