Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007039.hg.1 |
ENST00000378138.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007039.hg.1 |
ENST00000378138.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007039.hg.1 |
ENST00000378138.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007039.hg.1 |
ENST00000378138.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007039.hg.1 |
ENST00000378138.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007039.hg.1 |
ENST00000378138.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007039.hg.1 |
ENST00000378138.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007039.hg.1 |
ENST00000378142.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007039.hg.1 |
ENST00000378142.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007039.hg.1 |
ENST00000378142.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007039.hg.1 |
ENST00000378142.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007039.hg.1 |
ENST00000378142.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007039.hg.1 |
ENST00000378142.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007039.hg.1 |
ENST00000378142.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007039.hg.1 |
ENST00000378142.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007039.hg.1 |
ENST00000378142.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007039.hg.1 |
ENST00000620846.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007039.hg.1 |
ENST00000620846.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007039.hg.1 |
ENST00000620846.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007039.hg.1 |
ENST00000620846.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007039.hg.1 |
ENST00000620846.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007039.hg.1 |
NM_001097579.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007039.hg.1 |
NM_001097579.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007039.hg.1 |
NM_001097579.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007039.hg.1 |
NM_001097579.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007039.hg.1 |
NM_001097579.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007039.hg.1 |
NM_001097579.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007039.hg.1 |
NM_001097579.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007039.hg.1 |
NM_001097579.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007039.hg.1 |
NM_001097579.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007039.hg.1 |
NM_005300.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007039.hg.1 |
NM_005300.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007039.hg.1 |
NM_005300.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007039.hg.1 |
NM_005300.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007039.hg.1 |
NM_005300.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007039.hg.1 |
NM_005300.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007039.hg.1 |
NM_005300.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007039.hg.1 |
NM_005300.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X002257.hg.1 |
TC0X000202.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X002257.hg.1 |
ENST00000378142.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X002257.hg.1 |
ENST00000535368.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X002257.hg.1 |
NM_001097579.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007039.hg.1 |
ENST00000378138.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007039.hg.1 |
ENST00000378138.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007039.hg.1 |
ENST00000378138.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007039.hg.1 |
ENST00000378138.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007039.hg.1 |
ENST00000378138.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007039.hg.1 |
ENST00000378138.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007039.hg.1 |
ENST00000378138.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007039.hg.1 |
ENST00000378142.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007039.hg.1 |
ENST00000378142.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007039.hg.1 |
ENST00000378142.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007039.hg.1 |
ENST00000378142.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007039.hg.1 |
ENST00000378142.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007039.hg.1 |
ENST00000378142.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007039.hg.1 |
ENST00000378142.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007039.hg.1 |
ENST00000378142.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007039.hg.1 |
ENST00000378142.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007039.hg.1 |
ENST00000620846.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007039.hg.1 |
ENST00000620846.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007039.hg.1 |
ENST00000620846.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007039.hg.1 |
ENST00000620846.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007039.hg.1 |
ENST00000620846.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007039.hg.1 |
NM_001097579.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007039.hg.1 |
NM_001097579.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007039.hg.1 |
NM_001097579.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007039.hg.1 |
NM_001097579.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007039.hg.1 |
NM_001097579.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007039.hg.1 |
NM_001097579.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007039.hg.1 |
NM_001097579.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007039.hg.1 |
NM_001097579.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007039.hg.1 |
NM_001097579.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007039.hg.1 |
NM_005300.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007039.hg.1 |
NM_005300.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007039.hg.1 |
NM_005300.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007039.hg.1 |
NM_005300.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007039.hg.1 |
NM_005300.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007039.hg.1 |
NM_005300.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007039.hg.1 |
NM_005300.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007039.hg.1 |
NM_005300.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00147928.hg.1 |
ENST00000378142.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00147928.hg.1 |
ENST00000620846.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00147928.hg.1 |
NM_001097579.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00147928.hg.1 |
ENST00000378142.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00147928.hg.1 |
ENST00000620846.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00147928.hg.1 |
NM_001097579.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00147928.hg.1 |
TC0X00007039.hg.1 |
Clariom D TC |