Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012341.hg.1 |
ENST00000225698.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012341.hg.1 |
ENST00000225698.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012341.hg.1 |
ENST00000225698.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012341.hg.1 |
ENST00000225698.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012341.hg.1 |
ENST00000225698.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012341.hg.1 |
ENST00000225698.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012341.hg.1 |
ENST00000225698.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012341.hg.1 |
ENST00000225698.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012341.hg.1 |
ENST00000225698.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012341.hg.1 |
ENST00000225698.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012341.hg.1 |
ENST00000225698.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012341.hg.1 |
ENST00000573204.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012341.hg.1 |
ENST00000573204.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012341.hg.1 |
ENST00000573204.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012341.hg.1 |
ENST00000573204.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012341.hg.1 |
ENST00000573204.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012341.hg.1 |
ENST00000573204.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012341.hg.1 |
ENST00000573406.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012341.hg.1 |
ENST00000573406.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012341.hg.1 |
ENST00000573406.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012341.hg.1 |
ENST00000573406.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012341.hg.1 |
ENST00000573406.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012341.hg.1 |
ENST00000573406.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012341.hg.1 |
ENST00000573421.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012341.hg.1 |
ENST00000573421.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012341.hg.1 |
ENST00000573421.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012341.hg.1 |
ENST00000574444.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012341.hg.1 |
ENST00000574444.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012341.hg.1 |
ENST00000574444.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012341.hg.1 |
ENST00000574444.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012341.hg.1 |
ENST00000574444.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012341.hg.1 |
ENST00000574444.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012341.hg.1 |
ENST00000574444.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012341.hg.1 |
ENST00000574444.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012341.hg.1 |
ENST00000574444.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012341.hg.1 |
ENST00000574444.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012341.hg.1 |
ENST00000575890.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012341.hg.1 |
ENST00000575890.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012341.hg.1 |
ENST00000575890.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012341.hg.1 |
ENST00000575890.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012341.hg.1 |
ENST00000576122.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012341.hg.1 |
ENST00000576122.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012341.hg.1 |
ENST00000576122.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012341.hg.1 |
NM_001212.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012341.hg.1 |
NM_001212.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012341.hg.1 |
NM_001212.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012341.hg.1 |
NM_001212.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012341.hg.1 |
NM_001212.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012341.hg.1 |
NM_001212.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012341.hg.1 |
NM_001212.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012341.hg.1 |
NM_001212.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012341.hg.1 |
NM_001212.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012341.hg.1 |
NM_001212.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012341.hg.1 |
NM_001212.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17014125.hg.1 |
ENST00000225698.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17014125.hg.1 |
NM_001212.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17014125.hg.1 |
TC17001056.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1700204865.hg.1 |
ENST00000225698.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700204865.hg.1 |
ENST00000573204.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700204865.hg.1 |
ENST00000573421.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700204865.hg.1 |
ENST00000574444.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700204865.hg.1 |
NM_001212.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700204865.hg.1 |
ENST00000225698.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700204865.hg.1 |
ENST00000573204.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700204865.hg.1 |
ENST00000573421.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700204865.hg.1 |
ENST00000574444.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700204865.hg.1 |
NM_001212.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700204865.hg.1 |
TC1700012341.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1700012341.hg.1 |
ENST00000225698.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012341.hg.1 |
ENST00000225698.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012341.hg.1 |
ENST00000225698.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012341.hg.1 |
ENST00000225698.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012341.hg.1 |
ENST00000225698.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012341.hg.1 |
ENST00000225698.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012341.hg.1 |
ENST00000225698.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012341.hg.1 |
ENST00000225698.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012341.hg.1 |
ENST00000225698.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012341.hg.1 |
ENST00000225698.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012341.hg.1 |
ENST00000225698.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012341.hg.1 |
ENST00000573204.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012341.hg.1 |
ENST00000573204.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012341.hg.1 |
ENST00000573204.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012341.hg.1 |
ENST00000573204.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012341.hg.1 |
ENST00000573204.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012341.hg.1 |
ENST00000573204.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012341.hg.1 |
ENST00000573406.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012341.hg.1 |
ENST00000573406.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012341.hg.1 |
ENST00000573406.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012341.hg.1 |
ENST00000573406.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012341.hg.1 |
ENST00000573406.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012341.hg.1 |
ENST00000573406.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012341.hg.1 |
ENST00000573421.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012341.hg.1 |
ENST00000573421.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012341.hg.1 |
ENST00000573421.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012341.hg.1 |
ENST00000574444.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012341.hg.1 |
ENST00000574444.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012341.hg.1 |
ENST00000574444.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012341.hg.1 |
ENST00000574444.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012341.hg.1 |
ENST00000574444.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012341.hg.1 |
ENST00000574444.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012341.hg.1 |
ENST00000574444.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012341.hg.1 |
ENST00000574444.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012341.hg.1 |
ENST00000574444.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012341.hg.1 |
ENST00000574444.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012341.hg.1 |
ENST00000575890.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012341.hg.1 |
ENST00000575890.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012341.hg.1 |
ENST00000575890.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012341.hg.1 |
ENST00000575890.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012341.hg.1 |
ENST00000576122.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012341.hg.1 |
ENST00000576122.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012341.hg.1 |
ENST00000576122.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012341.hg.1 |
NM_001212.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012341.hg.1 |
NM_001212.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012341.hg.1 |
NM_001212.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012341.hg.1 |
NM_001212.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012341.hg.1 |
NM_001212.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012341.hg.1 |
NM_001212.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012341.hg.1 |
NM_001212.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012341.hg.1 |
NM_001212.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012341.hg.1 |
NM_001212.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012341.hg.1 |
NM_001212.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012341.hg.1 |
NM_001212.3 |
Refseq |