Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZDHHC15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144969.2 | NP_659406.1 | 2 - 3 | 633 | 75 | |
XM_006724624.3 | XP_006724687.1 | 2 - 3 | 170 | 75 | ||
XM_017029296.1 | XP_016884785.1 | 2 - 3 | 170 | 75 | ||
XM_017029297.1 | XP_016884786.1 | 2 - 3 | 176 | 75 | ||
XM_017029298.1 | XP_016884787.1 | 2 - 3 | 164 | 75 | ||
GenBank mRNA | AK056374.1 | - | 2 - 3 | 171 | 75 | |
BC103980.1 | - | 2 - 3 | 171 | 40016292 | 75 | |
BC103981.1 | - | 2 - 3 | 171 | 40016293 | 75 | |
BC103982.1 | - | 2 - 3 | 171 | 40016294 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons |