Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM20C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020223.3 | NP_064608.2 | 7 - 8 | 1594 | 131 | |
XM_017012450.1 | XP_016867939.1 | 8 - 9 | 1850 | 131 | ||
XM_017012451.1 | XP_016867940.1 | 8 - 9 | 1847 | 131 | ||
XM_017012455.1 | XP_016867944.1 | 6 - 7 | 4433 | 131 | ||
GenBank mRNA | AB545605.1 | - | 7 - 8 | 1594 | 131 | |
AF533706.1 | - | 7 - 8 | 1594 | 131 | ||
AK054704.1 | - | 4 - 5 | 464 | 131 | ||
AL390147.1 | - | 4 - 5 | 1020 | 131 | ||
BC040074.1 | - | 7 - 8 | 1450 | 4942737 | 131 | |
BC087853.1 | - | 4 - 5 | 485 | 131 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |