Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM20C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017012450.1 | XP_016867939.1 | 2 - 3 | 1011 | 54 | |
XM_017012452.1 | XP_016867941.1 | 2 - 3 | 1011 | 54 | ||
XM_017012453.1 | XP_016867942.1 | 2 - 3 | 1046 | 54 | ||
XM_017012454.1 | XP_016867943.1 | 2 - 3 | 1026 | 54 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Homo sapiens Probe spans exons |