Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTMR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021090.3 | NP_066576.1 | 19 - 20 | 3751 | 63 | |
NM_153051.2 | NP_694691.1 | 18 - 19 | 3640 | 63 | ||
XM_005261805.2 | XP_005261862.1 | 20 - 21 | 3869 | 63 | ||
XM_005261808.2 | XP_005261865.1 | 19 - 20 | 3758 | 63 | ||
XM_017029029.1 | XP_016884518.1 | 18 - 19 | 3633 | 63 | ||
XM_017029031.1 | XP_016884520.1 | 17 - 18 | 3526 | 63 | ||
XM_017029032.1 | XP_016884521.1 | 18 - 19 | 3523 | 63 | ||
GenBank mRNA | AB002369.1 | - | 19 - 20 | 3670 | 63 | |
AF233437.1 | - | 15 - 16 | 3321 | 63 | ||
AF233438.1 | - | 16 - 17 | 3432 | 63 | ||
BC142713.1 | - | 18 - 19 | 3596 | 40148839 | 63 | |
BC148216.1 | - | 17 - 18 | 3459 | 40082266 | 63 | |
BC151217.1 | - | 19 - 20 | 3765 | 40148098 | 63 | |
BC152455.1 | - | 19 - 20 | 3670 | 8862588 | 63 | |
CR456525.1 | - | 17 - 18 | 3447 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Phosphatase 3' Most Mammalian Gene Collection Homo sapiens Probe spans exons |