Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR04010721.hg.1 |
TC04000777.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04010721.hg.1 |
ENST00000302068.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04010721.hg.1 |
ENST00000502545.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04010721.hg.1 |
ENST00000537843.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04010721.hg.1 |
NM_001184741.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04010721.hg.1 |
NM_005141.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400162168.hg.1 |
ENST00000302068.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400162168.hg.1 |
ENST00000425838.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400162168.hg.1 |
ENST00000473984.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400162168.hg.1 |
ENST00000497097.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400162168.hg.1 |
ENST00000502545.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400162168.hg.1 |
NM_001184741.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400162168.hg.1 |
NM_005141.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400162168.hg.1 |
ENST00000302068.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400162168.hg.1 |
ENST00000425838.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400162168.hg.1 |
ENST00000473984.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400162168.hg.1 |
ENST00000497097.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400162168.hg.1 |
ENST00000502545.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400162168.hg.1 |
NM_001184741.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400162168.hg.1 |
NM_005141.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400162168.hg.1 |
TC0400009064.hg.1 |
Clariom D TC |