Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C12orf40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_135051.1 | 9 - 10 | 1506 | 127 | ||
GenBank mRNA | AK097445.1 | - | 9 - 10 | 1455 | 127 | |
EU668334.1 | - | 9 - 10 | 1455 | 127 | ||
GQ472216.1 | - | 9 - 10 | 1455 | 127 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | ATP binding cassette subfamily D member 2 | ABCD2 |
ABC39, ALDL1, ALDR, ALDRP, hALDR |
-32.8kb | Anti-sense | Chr. 12 - 39530966 - 39651333 on NCBI Build GRCh38 |
Downstream | solute carrier family 2 member 13 | SLC2A13 | HMIT | 70.1kb | Anti-sense | Chr. 12 - 39755021 - 40106085 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Probe spans exons |