Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR19003856.hg.1 |
ENST00000269703.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19003856.hg.1 |
NM_173483.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19003856.hg.1 |
TC19000272.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007233.hg.1 |
ENST00000269703.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007233.hg.1 |
ENST00000269703.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007233.hg.1 |
ENST00000269703.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007233.hg.1 |
ENST00000269703.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007233.hg.1 |
ENST00000269703.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007233.hg.1 |
ENST00000269703.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007233.hg.1 |
ENST00000269703.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007233.hg.1 |
ENST00000269703.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007233.hg.1 |
ENST00000269703.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007233.hg.1 |
ENST00000269703.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007233.hg.1 |
ENST00000269703.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007233.hg.1 |
ENST00000269703.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007233.hg.1 |
ENST00000269703.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007233.hg.1 |
ENST00000269703.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007233.hg.1 |
ENST00000269703.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007233.hg.1 |
ENST00000269703.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007233.hg.1 |
ENST00000269703.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007233.hg.1 |
ENST00000269703.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007233.hg.1 |
ENST00000269703.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007233.hg.1 |
ENST00000601005.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007233.hg.1 |
ENST00000601005.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007233.hg.1 |
ENST00000601005.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007233.hg.1 |
ENST00000601005.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007233.hg.1 |
ENST00000601005.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007233.hg.1 |
ENST00000601005.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007233.hg.1 |
ENST00000601005.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007233.hg.1 |
ENST00000601005.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007233.hg.1 |
ENST00000601005.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007233.hg.1 |
ENST00000601005.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007233.hg.1 |
ENST00000601005.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007233.hg.1 |
ENST00000601005.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007233.hg.1 |
ENST00000601005.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007233.hg.1 |
ENST00000601005.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007233.hg.1 |
ENST00000601005.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007233.hg.1 |
NM_173483.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007233.hg.1 |
NM_173483.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007233.hg.1 |
NM_173483.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007233.hg.1 |
NM_173483.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007233.hg.1 |
NM_173483.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007233.hg.1 |
NM_173483.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007233.hg.1 |
NM_173483.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007233.hg.1 |
NM_173483.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007233.hg.1 |
NM_173483.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007233.hg.1 |
NM_173483.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007233.hg.1 |
NM_173483.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007233.hg.1 |
NM_173483.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007233.hg.1 |
NM_173483.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007233.hg.1 |
NM_173483.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007233.hg.1 |
NM_173483.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007233.hg.1 |
NM_173483.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007233.hg.1 |
NM_173483.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007233.hg.1 |
NM_173483.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900152147.hg.1 |
ENST00000269703.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900152147.hg.1 |
ENST00000601005.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900152147.hg.1 |
NM_173483.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900152147.hg.1 |
TC1900007233.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900152147.hg.1 |
ENST00000269703.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900152147.hg.1 |
ENST00000601005.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900152147.hg.1 |
NM_173483.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007233.hg.1 |
ENST00000269703.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007233.hg.1 |
ENST00000269703.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007233.hg.1 |
ENST00000269703.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007233.hg.1 |
ENST00000269703.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007233.hg.1 |
ENST00000269703.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007233.hg.1 |
ENST00000269703.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007233.hg.1 |
ENST00000269703.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007233.hg.1 |
ENST00000269703.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007233.hg.1 |
ENST00000269703.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007233.hg.1 |
ENST00000269703.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007233.hg.1 |
ENST00000269703.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007233.hg.1 |
ENST00000269703.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007233.hg.1 |
ENST00000269703.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007233.hg.1 |
ENST00000269703.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007233.hg.1 |
ENST00000269703.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007233.hg.1 |
ENST00000269703.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007233.hg.1 |
ENST00000269703.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007233.hg.1 |
ENST00000269703.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007233.hg.1 |
ENST00000269703.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007233.hg.1 |
ENST00000601005.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007233.hg.1 |
ENST00000601005.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007233.hg.1 |
ENST00000601005.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007233.hg.1 |
ENST00000601005.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007233.hg.1 |
ENST00000601005.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007233.hg.1 |
ENST00000601005.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007233.hg.1 |
ENST00000601005.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007233.hg.1 |
ENST00000601005.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007233.hg.1 |
ENST00000601005.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007233.hg.1 |
ENST00000601005.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007233.hg.1 |
ENST00000601005.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007233.hg.1 |
ENST00000601005.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007233.hg.1 |
ENST00000601005.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007233.hg.1 |
ENST00000601005.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007233.hg.1 |
ENST00000601005.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007233.hg.1 |
NM_173483.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007233.hg.1 |
NM_173483.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007233.hg.1 |
NM_173483.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007233.hg.1 |
NM_173483.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007233.hg.1 |
NM_173483.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007233.hg.1 |
NM_173483.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007233.hg.1 |
NM_173483.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007233.hg.1 |
NM_173483.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007233.hg.1 |
NM_173483.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007233.hg.1 |
NM_173483.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007233.hg.1 |
NM_173483.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007233.hg.1 |
NM_173483.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007233.hg.1 |
NM_173483.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007233.hg.1 |
NM_173483.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007233.hg.1 |
NM_173483.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007233.hg.1 |
NM_173483.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007233.hg.1 |
NM_173483.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007233.hg.1 |
NM_173483.3 |
Refseq |