Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR04005737.hg.1 |
TC04000414.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04005737.hg.1 |
ENST00000244869.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04005737.hg.1 |
NM_001432.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007853.hg.1 |
ENST00000244869.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007853.hg.1 |
ENST00000244869.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007853.hg.1 |
ENST00000244869.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007853.hg.1 |
ENST00000244869.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007853.hg.1 |
ENST00000244869.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007853.hg.1 |
ENST00000244869.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007853.hg.1 |
ENST00000244869.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007853.hg.1 |
ENST00000244869.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007853.hg.1 |
ENST00000244869.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007853.hg.1 |
ENST00000244869.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007853.hg.1 |
ENST00000503689.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007853.hg.1 |
ENST00000503689.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007853.hg.1 |
ENST00000503689.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007853.hg.1 |
ENST00000503689.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007853.hg.1 |
ENST00000507603.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007853.hg.1 |
ENST00000507603.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007853.hg.1 |
ENST00000507603.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007853.hg.1 |
ENST00000507603.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007853.hg.1 |
ENST00000507603.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007853.hg.1 |
NM_001432.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007853.hg.1 |
NM_001432.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007853.hg.1 |
NM_001432.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007853.hg.1 |
NM_001432.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007853.hg.1 |
NM_001432.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007853.hg.1 |
NM_001432.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007853.hg.1 |
NM_001432.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007853.hg.1 |
NM_001432.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007853.hg.1 |
NM_001432.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400007853.hg.1 |
NM_001432.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007853.hg.1 |
ENST00000244869.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007853.hg.1 |
ENST00000244869.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007853.hg.1 |
ENST00000244869.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007853.hg.1 |
ENST00000244869.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007853.hg.1 |
ENST00000244869.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007853.hg.1 |
ENST00000244869.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007853.hg.1 |
ENST00000244869.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007853.hg.1 |
ENST00000244869.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007853.hg.1 |
ENST00000244869.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007853.hg.1 |
ENST00000244869.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007853.hg.1 |
ENST00000503689.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007853.hg.1 |
ENST00000503689.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007853.hg.1 |
ENST00000503689.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007853.hg.1 |
ENST00000503689.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007853.hg.1 |
ENST00000507603.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007853.hg.1 |
ENST00000507603.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007853.hg.1 |
ENST00000507603.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007853.hg.1 |
ENST00000507603.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007853.hg.1 |
ENST00000507603.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400007853.hg.1 |
NM_001432.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007853.hg.1 |
NM_001432.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007853.hg.1 |
NM_001432.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007853.hg.1 |
NM_001432.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007853.hg.1 |
NM_001432.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007853.hg.1 |
NM_001432.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007853.hg.1 |
NM_001432.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007853.hg.1 |
NM_001432.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007853.hg.1 |
NM_001432.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400007853.hg.1 |
NM_001432.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400153861.hg.1 |
ENST00000244869.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400153861.hg.1 |
ENST00000507603.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400153861.hg.1 |
NM_001432.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400153861.hg.1 |
ENST00000244869.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400153861.hg.1 |
ENST00000507603.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400153861.hg.1 |
NM_001432.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400153861.hg.1 |
TC0400007853.hg.1 |
Clariom D TC |