Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR19023513.hg.1 |
ENST00000246801.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19023513.hg.1 |
NM_021733.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19023513.hg.1 |
TC19001723.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR19023512.hg.1 |
ENST00000246801.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19023512.hg.1 |
NM_021733.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19023512.hg.1 |
TC19001723.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011160.hg.1 |
ENST00000246801.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011160.hg.1 |
ENST00000246801.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011160.hg.1 |
ENST00000246801.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011160.hg.1 |
ENST00000246801.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011160.hg.1 |
ENST00000246801.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011160.hg.1 |
ENST00000246801.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011160.hg.1 |
ENST00000246801.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011160.hg.1 |
ENST00000246801.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011160.hg.1 |
ENST00000246801.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011160.hg.1 |
ENST00000246801.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011160.hg.1 |
ENST00000246801.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011160.hg.1 |
ENST00000246801.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011160.hg.1 |
ENST00000246801.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011160.hg.1 |
ENST00000358830.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011160.hg.1 |
ENST00000358830.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011160.hg.1 |
ENST00000358830.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011160.hg.1 |
ENST00000358830.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011160.hg.1 |
ENST00000358830.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011160.hg.1 |
ENST00000358830.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011160.hg.1 |
ENST00000358830.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011160.hg.1 |
ENST00000599325.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011160.hg.1 |
ENST00000599325.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011160.hg.1 |
ENST00000599325.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011160.hg.1 |
ENST00000599325.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011160.hg.1 |
NM_021733.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011160.hg.1 |
NM_021733.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011160.hg.1 |
NM_021733.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011160.hg.1 |
NM_021733.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011160.hg.1 |
NM_021733.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011160.hg.1 |
NM_021733.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011160.hg.1 |
NM_021733.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011160.hg.1 |
NM_021733.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011160.hg.1 |
NM_021733.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011160.hg.1 |
NM_021733.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011160.hg.1 |
NM_021733.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900193523.hg.1 |
ENST00000246801.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900193523.hg.1 |
NM_021733.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900193523.hg.1 |
TC1900011160.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900193523.hg.1 |
ENST00000246801.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900193523.hg.1 |
NM_021733.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900193524.hg.1 |
ENST00000246801.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900193524.hg.1 |
NM_021733.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900193524.hg.1 |
TC1900011160.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900193524.hg.1 |
ENST00000246801.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900193524.hg.1 |
NM_021733.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011160.hg.1 |
ENST00000246801.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011160.hg.1 |
ENST00000246801.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011160.hg.1 |
ENST00000246801.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011160.hg.1 |
ENST00000246801.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011160.hg.1 |
ENST00000246801.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011160.hg.1 |
ENST00000246801.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011160.hg.1 |
ENST00000246801.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011160.hg.1 |
ENST00000246801.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011160.hg.1 |
ENST00000246801.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011160.hg.1 |
ENST00000246801.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011160.hg.1 |
ENST00000246801.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011160.hg.1 |
ENST00000246801.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011160.hg.1 |
ENST00000246801.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011160.hg.1 |
ENST00000358830.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011160.hg.1 |
ENST00000358830.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011160.hg.1 |
ENST00000358830.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011160.hg.1 |
ENST00000358830.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011160.hg.1 |
ENST00000358830.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011160.hg.1 |
ENST00000358830.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011160.hg.1 |
ENST00000358830.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011160.hg.1 |
ENST00000599325.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011160.hg.1 |
ENST00000599325.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011160.hg.1 |
ENST00000599325.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011160.hg.1 |
ENST00000599325.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011160.hg.1 |
NM_021733.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011160.hg.1 |
NM_021733.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011160.hg.1 |
NM_021733.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011160.hg.1 |
NM_021733.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011160.hg.1 |
NM_021733.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011160.hg.1 |
NM_021733.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011160.hg.1 |
NM_021733.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011160.hg.1 |
NM_021733.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011160.hg.1 |
NM_021733.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011160.hg.1 |
NM_021733.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011160.hg.1 |
NM_021733.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900193526.hg.1 |
ENST00000246801.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900193526.hg.1 |
TC1900011160.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900193526.hg.1 |
ENST00000246801.7 |
Ensembl |