Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR05018880.hg.1 |
TC05001345.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05018880.hg.1 |
ENST00000502271.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05018880.hg.1 |
ENST00000504924.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05018880.hg.1 |
NM_019087.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010731.hg.1 |
ENST00000502271.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010731.hg.1 |
ENST00000502271.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010731.hg.1 |
ENST00000502271.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010731.hg.1 |
ENST00000502271.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010731.hg.1 |
ENST00000502271.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010731.hg.1 |
ENST00000502271.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010731.hg.1 |
ENST00000502271.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010731.hg.1 |
ENST00000502271.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010731.hg.1 |
ENST00000502271.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010731.hg.1 |
ENST00000502271.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010731.hg.1 |
ENST00000504924.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010731.hg.1 |
ENST00000504924.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010731.hg.1 |
ENST00000504924.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010731.hg.1 |
ENST00000504924.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010731.hg.1 |
ENST00000504924.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010731.hg.1 |
ENST00000504924.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010731.hg.1 |
ENST00000504924.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010731.hg.1 |
ENST00000504924.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010731.hg.1 |
ENST00000504924.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010731.hg.1 |
ENST00000505383.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010731.hg.1 |
ENST00000505383.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010731.hg.1 |
ENST00000505383.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010731.hg.1 |
ENST00000505383.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010731.hg.1 |
ENST00000505383.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010731.hg.1 |
ENST00000505630.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010731.hg.1 |
ENST00000505630.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010731.hg.1 |
ENST00000505630.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010731.hg.1 |
ENST00000505630.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010731.hg.1 |
ENST00000505630.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010731.hg.1 |
ENST00000505630.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010731.hg.1 |
ENST00000507646.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010731.hg.1 |
ENST00000507646.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010731.hg.1 |
ENST00000507646.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010731.hg.1 |
ENST00000507646.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010731.hg.1 |
ENST00000507646.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010731.hg.1 |
ENST00000510591.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010731.hg.1 |
ENST00000510591.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010731.hg.1 |
ENST00000510591.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010731.hg.1 |
ENST00000510591.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010731.hg.1 |
ENST00000510591.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010731.hg.1 |
ENST00000510591.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010731.hg.1 |
ENST00000620747.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010731.hg.1 |
ENST00000620747.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010731.hg.1 |
ENST00000620747.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010731.hg.1 |
ENST00000620747.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010731.hg.1 |
ENST00000620747.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010731.hg.1 |
ENST00000620747.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010731.hg.1 |
ENST00000620747.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010731.hg.1 |
ENST00000620747.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010731.hg.1 |
ENST00000620747.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010731.hg.1 |
NM_019087.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010731.hg.1 |
NM_019087.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010731.hg.1 |
NM_019087.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010731.hg.1 |
NM_019087.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010731.hg.1 |
NM_019087.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010731.hg.1 |
NM_019087.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010731.hg.1 |
NM_019087.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010731.hg.1 |
NM_019087.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500010731.hg.1 |
NM_019087.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500174883.hg.1 |
ENST00000502271.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500174883.hg.1 |
ENST00000504924.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500174883.hg.1 |
ENST00000505630.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500174883.hg.1 |
ENST00000507646.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500174883.hg.1 |
ENST00000510591.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500174883.hg.1 |
ENST00000620747.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500174883.hg.1 |
NM_019087.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500174883.hg.1 |
ENST00000502271.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500174883.hg.1 |
ENST00000504924.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500174883.hg.1 |
ENST00000505630.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500174883.hg.1 |
ENST00000507646.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500174883.hg.1 |
ENST00000510591.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500174883.hg.1 |
ENST00000620747.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500174883.hg.1 |
NM_019087.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500174883.hg.1 |
TC0500010731.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0500010731.hg.1 |
ENST00000502271.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010731.hg.1 |
ENST00000502271.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010731.hg.1 |
ENST00000502271.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010731.hg.1 |
ENST00000502271.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010731.hg.1 |
ENST00000502271.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010731.hg.1 |
ENST00000502271.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010731.hg.1 |
ENST00000502271.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010731.hg.1 |
ENST00000502271.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010731.hg.1 |
ENST00000502271.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010731.hg.1 |
ENST00000502271.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010731.hg.1 |
ENST00000504924.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010731.hg.1 |
ENST00000504924.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010731.hg.1 |
ENST00000504924.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010731.hg.1 |
ENST00000504924.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010731.hg.1 |
ENST00000504924.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010731.hg.1 |
ENST00000504924.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010731.hg.1 |
ENST00000504924.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010731.hg.1 |
ENST00000504924.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010731.hg.1 |
ENST00000504924.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010731.hg.1 |
ENST00000505383.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010731.hg.1 |
ENST00000505383.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010731.hg.1 |
ENST00000505383.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010731.hg.1 |
ENST00000505383.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010731.hg.1 |
ENST00000505383.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010731.hg.1 |
ENST00000505630.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010731.hg.1 |
ENST00000505630.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010731.hg.1 |
ENST00000505630.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010731.hg.1 |
ENST00000505630.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010731.hg.1 |
ENST00000505630.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010731.hg.1 |
ENST00000505630.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010731.hg.1 |
ENST00000507646.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010731.hg.1 |
ENST00000507646.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010731.hg.1 |
ENST00000507646.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010731.hg.1 |
ENST00000507646.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010731.hg.1 |
ENST00000507646.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010731.hg.1 |
ENST00000510591.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010731.hg.1 |
ENST00000510591.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010731.hg.1 |
ENST00000510591.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010731.hg.1 |
ENST00000510591.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010731.hg.1 |
ENST00000510591.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010731.hg.1 |
ENST00000510591.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010731.hg.1 |
ENST00000620747.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010731.hg.1 |
ENST00000620747.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010731.hg.1 |
ENST00000620747.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010731.hg.1 |
ENST00000620747.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010731.hg.1 |
ENST00000620747.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010731.hg.1 |
ENST00000620747.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010731.hg.1 |
ENST00000620747.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010731.hg.1 |
ENST00000620747.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010731.hg.1 |
ENST00000620747.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500010731.hg.1 |
NM_019087.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010731.hg.1 |
NM_019087.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010731.hg.1 |
NM_019087.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010731.hg.1 |
NM_019087.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010731.hg.1 |
NM_019087.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010731.hg.1 |
NM_019087.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010731.hg.1 |
NM_019087.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010731.hg.1 |
NM_019087.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500010731.hg.1 |
NM_019087.2 |
Refseq |