Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013604.hg.1 |
ENST00000206262.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013604.hg.1 |
ENST00000206262.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013604.hg.1 |
ENST00000206262.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013604.hg.1 |
ENST00000206262.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013604.hg.1 |
ENST00000206262.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013604.hg.1 |
ENST00000206262.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013604.hg.1 |
ENST00000206262.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013604.hg.1 |
ENST00000206262.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013604.hg.1 |
ENST00000367225.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013604.hg.1 |
ENST00000367225.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013604.hg.1 |
ENST00000367225.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013604.hg.1 |
ENST00000367225.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013604.hg.1 |
ENST00000367225.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013604.hg.1 |
ENST00000367225.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013604.hg.1 |
ENST00000367225.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013604.hg.1 |
ENST00000367225.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013604.hg.1 |
NM_012419.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013604.hg.1 |
NM_012419.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013604.hg.1 |
NM_012419.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013604.hg.1 |
NM_012419.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013604.hg.1 |
NM_012419.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013604.hg.1 |
NM_012419.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013604.hg.1 |
NM_012419.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06028207.hg.1 |
TC06002242.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06028207.hg.1 |
ENST00000206262.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06028207.hg.1 |
ENST00000367225.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06028207.hg.1 |
NM_012419.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600194083.hg.1 |
ENST00000206262.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600194083.hg.1 |
ENST00000367225.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600194083.hg.1 |
NM_012419.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600194083.hg.1 |
ENST00000206262.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600194083.hg.1 |
ENST00000367225.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600194083.hg.1 |
NM_012419.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600194083.hg.1 |
TC0600013604.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600013604.hg.1 |
ENST00000206262.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013604.hg.1 |
ENST00000206262.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013604.hg.1 |
ENST00000206262.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013604.hg.1 |
ENST00000206262.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013604.hg.1 |
ENST00000206262.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013604.hg.1 |
ENST00000206262.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013604.hg.1 |
ENST00000206262.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013604.hg.1 |
ENST00000206262.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013604.hg.1 |
ENST00000367225.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013604.hg.1 |
ENST00000367225.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013604.hg.1 |
ENST00000367225.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013604.hg.1 |
ENST00000367225.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013604.hg.1 |
ENST00000367225.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013604.hg.1 |
ENST00000367225.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013604.hg.1 |
ENST00000367225.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013604.hg.1 |
ENST00000367225.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013604.hg.1 |
NM_012419.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013604.hg.1 |
NM_012419.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013604.hg.1 |
NM_012419.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013604.hg.1 |
NM_012419.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013604.hg.1 |
NM_012419.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013604.hg.1 |
NM_012419.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013604.hg.1 |
NM_012419.4 |
Refseq |