Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06008767.hg.1 |
TC06000733.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06008767.hg.1 |
ENST00000370336.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06008767.hg.1 |
ENST00000479773.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06008767.hg.1 |
NM_018665.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008527.hg.1 |
ENST00000370336.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008527.hg.1 |
ENST00000370336.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008527.hg.1 |
ENST00000370336.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008527.hg.1 |
ENST00000370336.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008527.hg.1 |
ENST00000370336.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008527.hg.1 |
ENST00000370336.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008527.hg.1 |
ENST00000370336.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008527.hg.1 |
ENST00000370336.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008527.hg.1 |
ENST00000370336.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008527.hg.1 |
ENST00000370336.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008527.hg.1 |
ENST00000370336.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008527.hg.1 |
ENST00000370336.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008527.hg.1 |
ENST00000370336.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008527.hg.1 |
ENST00000370336.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008527.hg.1 |
ENST00000370336.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008527.hg.1 |
ENST00000370336.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008527.hg.1 |
ENST00000370336.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008527.hg.1 |
ENST00000370336.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008527.hg.1 |
ENST00000370336.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008527.hg.1 |
ENST00000370336.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008527.hg.1 |
ENST00000370336.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008527.hg.1 |
ENST00000464221.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008527.hg.1 |
ENST00000464221.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008527.hg.1 |
ENST00000464221.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008527.hg.1 |
ENST00000479773.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008527.hg.1 |
ENST00000479773.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008527.hg.1 |
ENST00000479773.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008527.hg.1 |
ENST00000479773.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008527.hg.1 |
ENST00000479773.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008527.hg.1 |
ENST00000479773.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008527.hg.1 |
ENST00000479773.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008527.hg.1 |
NM_018665.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008527.hg.1 |
NM_018665.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008527.hg.1 |
NM_018665.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008527.hg.1 |
NM_018665.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008527.hg.1 |
NM_018665.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008527.hg.1 |
NM_018665.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008527.hg.1 |
NM_018665.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008527.hg.1 |
NM_018665.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008527.hg.1 |
NM_018665.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008527.hg.1 |
NM_018665.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008527.hg.1 |
NM_018665.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008527.hg.1 |
NM_018665.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008527.hg.1 |
NM_018665.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008527.hg.1 |
NM_018665.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008527.hg.1 |
NM_018665.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008527.hg.1 |
NM_018665.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008527.hg.1 |
NM_018665.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008527.hg.1 |
NM_018665.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008527.hg.1 |
NM_018665.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008527.hg.1 |
NM_018665.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008527.hg.1 |
NM_018665.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008527.hg.1 |
NM_018665.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600159266.hg.1 |
ENST00000370336.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600159266.hg.1 |
ENST00000479773.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600159266.hg.1 |
NM_018665.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600159266.hg.1 |
ENST00000370336.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600159266.hg.1 |
ENST00000479773.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600159266.hg.1 |
NM_018665.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600159266.hg.1 |
TC0600008527.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600008527.hg.1 |
ENST00000370336.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008527.hg.1 |
ENST00000370336.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008527.hg.1 |
ENST00000370336.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008527.hg.1 |
ENST00000370336.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008527.hg.1 |
ENST00000370336.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008527.hg.1 |
ENST00000370336.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008527.hg.1 |
ENST00000370336.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008527.hg.1 |
ENST00000370336.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008527.hg.1 |
ENST00000370336.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008527.hg.1 |
ENST00000370336.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008527.hg.1 |
ENST00000370336.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008527.hg.1 |
ENST00000370336.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008527.hg.1 |
ENST00000370336.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008527.hg.1 |
ENST00000370336.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008527.hg.1 |
ENST00000370336.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008527.hg.1 |
ENST00000370336.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008527.hg.1 |
ENST00000370336.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008527.hg.1 |
ENST00000370336.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008527.hg.1 |
ENST00000370336.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008527.hg.1 |
ENST00000370336.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008527.hg.1 |
ENST00000370336.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008527.hg.1 |
ENST00000464221.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008527.hg.1 |
ENST00000464221.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008527.hg.1 |
ENST00000464221.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008527.hg.1 |
ENST00000479773.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008527.hg.1 |
ENST00000479773.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008527.hg.1 |
ENST00000479773.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008527.hg.1 |
ENST00000479773.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008527.hg.1 |
ENST00000479773.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008527.hg.1 |
ENST00000479773.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008527.hg.1 |
ENST00000479773.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008527.hg.1 |
NM_018665.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008527.hg.1 |
NM_018665.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008527.hg.1 |
NM_018665.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008527.hg.1 |
NM_018665.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008527.hg.1 |
NM_018665.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008527.hg.1 |
NM_018665.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008527.hg.1 |
NM_018665.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008527.hg.1 |
NM_018665.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008527.hg.1 |
NM_018665.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008527.hg.1 |
NM_018665.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008527.hg.1 |
NM_018665.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008527.hg.1 |
NM_018665.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008527.hg.1 |
NM_018665.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008527.hg.1 |
NM_018665.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008527.hg.1 |
NM_018665.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008527.hg.1 |
NM_018665.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008527.hg.1 |
NM_018665.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008527.hg.1 |
NM_018665.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008527.hg.1 |
NM_018665.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008527.hg.1 |
NM_018665.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008527.hg.1 |
NM_018665.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008527.hg.1 |
NM_018665.2 |
Refseq |