Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL6A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_004369.3 | NP_004360.2 | 19 - 20 | 6639 | 86 | |
NM_057166.4 | NP_476507.3 | 16 - 17 | 4818 | 86 | ||
NM_057167.3 | NP_476508.2 | 18 - 19 | 6021 | 86 | ||
XM_005246065.1 | XP_005246122.1 | 18 - 19 | 5996 | 86 | ||
XM_005246066.1 | XP_005246123.1 | 16 - 17 | 5142 | 86 | ||
XM_006712253.1 | XP_006712316.1 | 19 - 20 | 6095 | 86 | ||
XM_011510574.1 | XP_011508876.1 | 19 - 20 | 6593 | 86 | ||
XM_017003303.1 | XP_016858792.1 | 24 - 25 | 6646 | 86 | ||
XM_017003304.1 | XP_016858793.1 | 21 - 22 | 4858 | 86 | ||
GenBank mRNA | AB209636.1 | - | 10 - 11 | 1930 | 86 | |
BC144595.1 | - | 16 - 17 | 4759 | 9053125 | 86 | |
BC150625.1 | - | 16 - 17 | 4759 | 9056995 | 86 | |
BC171790.1 | - | 16 - 17 | 4759 | 9054444 | 86 | |
M20778.1 | - | 5 - 6 | 243 | 86 | ||
M27449.1 | - | 2 - 3 | 81 | 86 | ||
X06196.1 | - | 2 - 3 | 81 | 86 | ||
X52022.1 | - | 19 - 20 | 6609 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |