Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL6A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_004369.3 | NP_004360.2 | 20 - 21 | 6694 | 113 | |
NM_057166.4 | NP_476507.3 | 17 - 18 | 4873 | 113 | ||
NM_057167.3 | NP_476508.2 | 19 - 20 | 6076 | 113 | ||
XM_005246065.1 | XP_005246122.1 | 19 - 20 | 6051 | 113 | ||
XM_005246066.1 | XP_005246123.1 | 17 - 18 | 5197 | 113 | ||
XM_006712253.1 | XP_006712316.1 | 20 - 21 | 6150 | 113 | ||
XM_011510574.1 | XP_011508876.1 | 20 - 21 | 6648 | 113 | ||
XM_017003303.1 | XP_016858792.1 | 25 - 26 | 6701 | 113 | ||
XM_017003304.1 | XP_016858793.1 | 22 - 23 | 4913 | 113 | ||
GenBank mRNA | AB209636.1 | - | 11 - 12 | 1985 | 113 | |
BC144595.1 | - | 17 - 18 | 4814 | 9053125 | 113 | |
BC150625.1 | - | 17 - 18 | 4814 | 9056995 | 113 | |
BC171790.1 | - | 17 - 18 | 4814 | 9054444 | 113 | |
M20778.1 | - | 6 - 7 | 298 | 113 | ||
X06196.1 | - | 3 - 4 | 136 | 113 | ||
X52022.1 | - | 20 - 21 | 6664 | 113 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |