Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
ENST00000359520.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
ENST00000359520.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
ENST00000359520.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
ENST00000359520.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
ENST00000359520.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
ENST00000359520.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
ENST00000359520.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
ENST00000359520.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
ENST00000359520.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
ENST00000359520.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
ENST00000359520.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
ENST00000359520.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
ENST00000359520.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
ENST00000359520.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
ENST00000359520.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
ENST00000359520.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
ENST00000359520.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
ENST00000359520.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
ENST00000359520.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
ENST00000359520.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
ENST00000359520.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
ENST00000359520.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
ENST00000359520.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
ENST00000359520.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
ENST00000359520.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
ENST00000359520.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
ENST00000359520.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
ENST00000359520.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
ENST00000359520.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
ENST00000359520.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
ENST00000359520.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
ENST00000359520.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
ENST00000359520.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
ENST00000359520.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
ENST00000359520.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
ENST00000391028.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
ENST00000557786.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
ENST00000557786.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
ENST00000557786.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
ENST00000557786.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
ENST00000557786.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
ENST00000558678.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
ENST00000558678.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
ENST00000558678.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
ENST00000558678.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
ENST00000558678.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
ENST00000558678.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
ENST00000558678.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
ENST00000558678.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
ENST00000558678.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
ENST00000558678.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
ENST00000558678.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
ENST00000558678.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
ENST00000558678.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
ENST00000558678.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
ENST00000558678.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
ENST00000558678.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
ENST00000558678.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
ENST00000558678.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
ENST00000558678.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
ENST00000558678.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
ENST00000558678.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
ENST00000558678.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
ENST00000558678.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
ENST00000558678.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
ENST00000558678.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
ENST00000559124.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
ENST00000559124.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
ENST00000559124.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
ENST00000559124.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
ENST00000559124.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
ENST00000560060.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
ENST00000560060.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
ENST00000560060.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
ENST00000560060.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
ENST00000560060.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
ENST00000561099.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
ENST00000561099.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
ENST00000561099.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
ENST00000561099.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
ENST00000561228.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
ENST00000561228.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
ENST00000561228.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
ENST00000561228.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
ENST00000561228.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
ENST00000561228.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
ENST00000561228.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
NM_001172631.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
NM_001172631.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
NM_001172631.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
NM_001172631.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
NM_001172631.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
NM_001172631.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
NM_001172631.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
NM_001172631.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
NM_001172631.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
NM_001172631.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
NM_001172631.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
NM_001172631.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
NM_001172631.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
NM_001172631.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
NM_001172631.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
NM_001172631.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
NM_001172631.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
NM_001172631.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
NM_001172631.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
NM_001172631.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
NM_001172631.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
NM_001172631.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
NM_001172631.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
NM_001172631.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
NM_001172631.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
NM_001172631.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
NM_014844.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
NM_014844.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
NM_014844.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
NM_014844.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
NM_014844.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
NM_014844.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
NM_014844.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
NM_014844.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
NM_014844.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
NM_014844.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
NM_014844.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
NM_014844.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
NM_014844.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
NM_014844.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
NM_014844.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
NM_014844.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
NM_014844.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
NM_014844.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
NM_014844.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
NM_014844.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
NM_014844.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
NM_014844.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
NM_014844.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
NM_014844.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
NM_014844.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
NM_014844.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
NM_014844.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
NM_014844.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
NM_014844.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
NM_014844.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
NM_014844.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
NM_014844.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
NM_014844.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
NM_014844.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008349.hg.1 |
NM_014844.4 |
Refseq |