Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06011981.hg.1 |
TC06001019.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06011981.hg.1 |
ENST00000318471.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06011981.hg.1 |
ENST00000541292.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06011981.hg.1 |
NM_000288.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009566.hg.1 |
ENST00000318471.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009566.hg.1 |
ENST00000318471.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009566.hg.1 |
ENST00000318471.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009566.hg.1 |
ENST00000318471.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009566.hg.1 |
ENST00000318471.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009566.hg.1 |
ENST00000318471.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009566.hg.1 |
ENST00000318471.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009566.hg.1 |
ENST00000318471.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009566.hg.1 |
ENST00000318471.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009566.hg.1 |
ENST00000318471.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009566.hg.1 |
ENST00000318471.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009566.hg.1 |
ENST00000318471.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009566.hg.1 |
ENST00000318471.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009566.hg.1 |
ENST00000318471.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009566.hg.1 |
ENST00000318471.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009566.hg.1 |
ENST00000367756.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009566.hg.1 |
ENST00000367756.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009566.hg.1 |
ENST00000367756.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009566.hg.1 |
ENST00000367756.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009566.hg.1 |
ENST00000367756.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009566.hg.1 |
ENST00000367756.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009566.hg.1 |
ENST00000541292.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009566.hg.1 |
ENST00000541292.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009566.hg.1 |
ENST00000541292.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009566.hg.1 |
ENST00000541292.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009566.hg.1 |
ENST00000541292.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009566.hg.1 |
ENST00000541292.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009566.hg.1 |
ENST00000541292.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009566.hg.1 |
ENST00000541292.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009566.hg.1 |
ENST00000541292.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009566.hg.1 |
ENST00000541292.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009566.hg.1 |
NM_000288.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009566.hg.1 |
NM_000288.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009566.hg.1 |
NM_000288.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009566.hg.1 |
NM_000288.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009566.hg.1 |
NM_000288.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009566.hg.1 |
NM_000288.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009566.hg.1 |
NM_000288.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009566.hg.1 |
NM_000288.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009566.hg.1 |
NM_000288.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009566.hg.1 |
NM_000288.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009566.hg.1 |
NM_000288.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009566.hg.1 |
NM_000288.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009566.hg.1 |
NM_000288.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009566.hg.1 |
NM_000288.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009566.hg.1 |
NM_000288.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600165608.hg.1 |
ENST00000318471.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600165608.hg.1 |
ENST00000541292.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600165608.hg.1 |
NM_000288.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600165608.hg.1 |
ENST00000318471.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600165608.hg.1 |
ENST00000541292.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600165608.hg.1 |
NM_000288.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600165608.hg.1 |
TC0600009566.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600009566.hg.1 |
ENST00000318471.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009566.hg.1 |
ENST00000318471.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009566.hg.1 |
ENST00000318471.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009566.hg.1 |
ENST00000318471.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009566.hg.1 |
ENST00000318471.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009566.hg.1 |
ENST00000318471.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009566.hg.1 |
ENST00000318471.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009566.hg.1 |
ENST00000318471.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009566.hg.1 |
ENST00000318471.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009566.hg.1 |
ENST00000318471.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009566.hg.1 |
ENST00000318471.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009566.hg.1 |
ENST00000318471.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009566.hg.1 |
ENST00000318471.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009566.hg.1 |
ENST00000318471.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009566.hg.1 |
ENST00000318471.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009566.hg.1 |
ENST00000367756.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009566.hg.1 |
ENST00000367756.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009566.hg.1 |
ENST00000367756.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009566.hg.1 |
ENST00000367756.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009566.hg.1 |
ENST00000367756.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009566.hg.1 |
ENST00000367756.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009566.hg.1 |
ENST00000541292.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009566.hg.1 |
ENST00000541292.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009566.hg.1 |
ENST00000541292.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009566.hg.1 |
ENST00000541292.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009566.hg.1 |
ENST00000541292.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009566.hg.1 |
ENST00000541292.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009566.hg.1 |
ENST00000541292.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009566.hg.1 |
ENST00000541292.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009566.hg.1 |
ENST00000541292.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009566.hg.1 |
ENST00000541292.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009566.hg.1 |
NM_000288.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009566.hg.1 |
NM_000288.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009566.hg.1 |
NM_000288.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009566.hg.1 |
NM_000288.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009566.hg.1 |
NM_000288.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009566.hg.1 |
NM_000288.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009566.hg.1 |
NM_000288.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009566.hg.1 |
NM_000288.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009566.hg.1 |
NM_000288.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009566.hg.1 |
NM_000288.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009566.hg.1 |
NM_000288.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009566.hg.1 |
NM_000288.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009566.hg.1 |
NM_000288.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009566.hg.1 |
NM_000288.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009566.hg.1 |
NM_000288.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600165609.hg.1 |
ENST00000541292.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600165609.hg.1 |
ENST00000541292.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600165609.hg.1 |
TC0600009566.hg.1 |
Clariom D TC |