Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06007761.hg.1 |
TC06000637.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06007761.hg.1 |
ENST00000244799.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06007761.hg.1 |
ENST00000371211.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06007761.hg.1 |
ENST00000393699.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06007761.hg.1 |
ENST00000489301.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06007761.hg.1 |
NM_181744.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06007761.hg.1 |
NR_033806.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06007762.hg.1 |
TC06000637.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06007762.hg.1 |
ENST00000244799.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06007762.hg.1 |
ENST00000371211.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06007762.hg.1 |
ENST00000393699.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06007762.hg.1 |
ENST00000489301.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06007762.hg.1 |
NM_181744.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06007762.hg.1 |
NR_033806.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008190.hg.1 |
ENST00000244799.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008190.hg.1 |
ENST00000244799.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008190.hg.1 |
ENST00000244799.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008190.hg.1 |
ENST00000244799.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008190.hg.1 |
ENST00000244799.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008190.hg.1 |
ENST00000244799.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008190.hg.1 |
ENST00000244799.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008190.hg.1 |
ENST00000244799.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008190.hg.1 |
ENST00000244799.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008190.hg.1 |
ENST00000244799.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008190.hg.1 |
ENST00000244799.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008190.hg.1 |
ENST00000244799.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008190.hg.1 |
ENST00000244799.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008190.hg.1 |
ENST00000371211.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008190.hg.1 |
ENST00000371211.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008190.hg.1 |
ENST00000371211.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008190.hg.1 |
ENST00000371211.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008190.hg.1 |
ENST00000371211.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008190.hg.1 |
ENST00000371211.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008190.hg.1 |
ENST00000371211.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008190.hg.1 |
ENST00000371211.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008190.hg.1 |
ENST00000371211.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008190.hg.1 |
ENST00000371211.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008190.hg.1 |
ENST00000371211.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008190.hg.1 |
ENST00000371211.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008190.hg.1 |
ENST00000371211.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008190.hg.1 |
ENST00000393699.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008190.hg.1 |
ENST00000393699.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008190.hg.1 |
ENST00000393699.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008190.hg.1 |
ENST00000393699.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008190.hg.1 |
ENST00000393699.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008190.hg.1 |
ENST00000393699.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008190.hg.1 |
ENST00000393699.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008190.hg.1 |
ENST00000393699.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008190.hg.1 |
ENST00000393699.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008190.hg.1 |
ENST00000393699.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008190.hg.1 |
ENST00000393699.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008190.hg.1 |
ENST00000489301.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008190.hg.1 |
ENST00000489301.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008190.hg.1 |
ENST00000489301.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008190.hg.1 |
ENST00000489301.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008190.hg.1 |
ENST00000489301.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008190.hg.1 |
ENST00000489301.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008190.hg.1 |
ENST00000489301.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008190.hg.1 |
ENST00000489301.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008190.hg.1 |
ENST00000489301.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008190.hg.1 |
ENST00000489301.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008190.hg.1 |
ENST00000489301.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008190.hg.1 |
ENST00000489301.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008190.hg.1 |
ENST00000489301.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008190.hg.1 |
ENST00000510695.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008190.hg.1 |
ENST00000510695.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008190.hg.1 |
ENST00000510695.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008190.hg.1 |
ENST00000510695.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008190.hg.1 |
NM_181744.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008190.hg.1 |
NM_181744.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008190.hg.1 |
NM_181744.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008190.hg.1 |
NM_181744.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008190.hg.1 |
NM_181744.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008190.hg.1 |
NM_181744.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008190.hg.1 |
NM_181744.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008190.hg.1 |
NM_181744.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008190.hg.1 |
NM_181744.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008190.hg.1 |
NM_181744.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008190.hg.1 |
NM_181744.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008190.hg.1 |
NM_181744.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008190.hg.1 |
NM_181744.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008190.hg.1 |
NM_181744.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008190.hg.1 |
NR_033806.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008190.hg.1 |
NR_033806.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008190.hg.1 |
NR_033806.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008190.hg.1 |
NR_033806.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008190.hg.1 |
NR_033806.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008190.hg.1 |
NR_033806.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008190.hg.1 |
NR_033806.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008190.hg.1 |
NR_033806.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008190.hg.1 |
NR_033806.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008190.hg.1 |
NR_033806.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008190.hg.1 |
NR_033806.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008190.hg.1 |
NR_033806.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008190.hg.1 |
NR_033806.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600157320.hg.1 |
ENST00000244799.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600157320.hg.1 |
ENST00000371211.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600157320.hg.1 |
ENST00000393699.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600157320.hg.1 |
ENST00000489301.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600157320.hg.1 |
NM_181744.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600157320.hg.1 |
NR_033806.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600157320.hg.1 |
ENST00000244799.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600157320.hg.1 |
ENST00000371211.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600157320.hg.1 |
ENST00000393699.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600157320.hg.1 |
ENST00000489301.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600157320.hg.1 |
NM_181744.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600157320.hg.1 |
NR_033806.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600157320.hg.1 |
TC0600008190.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600008190.hg.1 |
ENST00000244799.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008190.hg.1 |
ENST00000244799.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008190.hg.1 |
ENST00000244799.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008190.hg.1 |
ENST00000244799.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008190.hg.1 |
ENST00000244799.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008190.hg.1 |
ENST00000244799.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008190.hg.1 |
ENST00000244799.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008190.hg.1 |
ENST00000244799.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008190.hg.1 |
ENST00000244799.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008190.hg.1 |
ENST00000244799.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008190.hg.1 |
ENST00000244799.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008190.hg.1 |
ENST00000244799.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008190.hg.1 |
ENST00000244799.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008190.hg.1 |
ENST00000371211.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008190.hg.1 |
ENST00000371211.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008190.hg.1 |
ENST00000371211.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008190.hg.1 |
ENST00000371211.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008190.hg.1 |
ENST00000371211.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008190.hg.1 |
ENST00000371211.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008190.hg.1 |
ENST00000371211.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008190.hg.1 |
ENST00000371211.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008190.hg.1 |
ENST00000371211.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008190.hg.1 |
ENST00000371211.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008190.hg.1 |
ENST00000371211.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008190.hg.1 |
ENST00000371211.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008190.hg.1 |
ENST00000371211.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008190.hg.1 |
ENST00000393699.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008190.hg.1 |
ENST00000393699.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008190.hg.1 |
ENST00000393699.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008190.hg.1 |
ENST00000393699.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008190.hg.1 |
ENST00000393699.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008190.hg.1 |
ENST00000393699.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008190.hg.1 |
ENST00000393699.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008190.hg.1 |
ENST00000393699.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008190.hg.1 |
ENST00000393699.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008190.hg.1 |
ENST00000393699.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008190.hg.1 |
ENST00000393699.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008190.hg.1 |
ENST00000489301.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008190.hg.1 |
ENST00000489301.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008190.hg.1 |
ENST00000489301.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008190.hg.1 |
ENST00000489301.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008190.hg.1 |
ENST00000489301.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008190.hg.1 |
ENST00000489301.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008190.hg.1 |
ENST00000489301.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008190.hg.1 |
ENST00000489301.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008190.hg.1 |
ENST00000489301.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008190.hg.1 |
ENST00000489301.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008190.hg.1 |
ENST00000489301.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008190.hg.1 |
ENST00000489301.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008190.hg.1 |
ENST00000489301.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008190.hg.1 |
ENST00000510695.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008190.hg.1 |
ENST00000510695.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008190.hg.1 |
ENST00000510695.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008190.hg.1 |
ENST00000510695.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008190.hg.1 |
NM_181744.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008190.hg.1 |
NM_181744.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008190.hg.1 |
NM_181744.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008190.hg.1 |
NM_181744.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008190.hg.1 |
NM_181744.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008190.hg.1 |
NM_181744.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008190.hg.1 |
NM_181744.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008190.hg.1 |
NM_181744.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008190.hg.1 |
NM_181744.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008190.hg.1 |
NM_181744.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008190.hg.1 |
NM_181744.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008190.hg.1 |
NM_181744.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008190.hg.1 |
NM_181744.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008190.hg.1 |
NM_181744.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008190.hg.1 |
NR_033806.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008190.hg.1 |
NR_033806.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008190.hg.1 |
NR_033806.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008190.hg.1 |
NR_033806.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008190.hg.1 |
NR_033806.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008190.hg.1 |
NR_033806.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008190.hg.1 |
NR_033806.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008190.hg.1 |
NR_033806.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008190.hg.1 |
NR_033806.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008190.hg.1 |
NR_033806.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008190.hg.1 |
NR_033806.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008190.hg.1 |
NR_033806.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008190.hg.1 |
NR_033806.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600157326.hg.1 |
ENST00000244799.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600157326.hg.1 |
ENST00000371211.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600157326.hg.1 |
ENST00000393699.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600157326.hg.1 |
ENST00000489301.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600157326.hg.1 |
NM_181744.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600157326.hg.1 |
NR_033806.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600157326.hg.1 |
ENST00000244799.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600157326.hg.1 |
ENST00000371211.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600157326.hg.1 |
ENST00000393699.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600157326.hg.1 |
ENST00000489301.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600157326.hg.1 |
NM_181744.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600157326.hg.1 |
NR_033806.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600157326.hg.1 |
TC0600008190.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600157325.hg.1 |
ENST00000244799.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600157325.hg.1 |
ENST00000371211.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600157325.hg.1 |
ENST00000393699.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600157325.hg.1 |
ENST00000489301.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600157325.hg.1 |
ENST00000510695.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600157325.hg.1 |
NM_181744.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600157325.hg.1 |
NR_033806.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600157325.hg.1 |
ENST00000244799.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600157325.hg.1 |
ENST00000371211.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600157325.hg.1 |
ENST00000393699.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600157325.hg.1 |
ENST00000489301.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600157325.hg.1 |
ENST00000510695.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600157325.hg.1 |
NM_181744.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600157325.hg.1 |
NR_033806.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600157325.hg.1 |
TC0600008190.hg.1 |
Clariom D TC |