Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008320.hg.1 |
ENST00000334077.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008320.hg.1 |
ENST00000334077.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008320.hg.1 |
ENST00000334077.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008320.hg.1 |
NM_005421.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008320.hg.1 |
NM_005421.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900008320.hg.1 |
NM_005421.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008320.hg.1 |
ENST00000334077.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008320.hg.1 |
ENST00000334077.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008320.hg.1 |
ENST00000334077.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900008320.hg.1 |
NM_005421.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008320.hg.1 |
NM_005421.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900008320.hg.1 |
NM_005421.2 |
Refseq |