Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007843.hg.1 |
ENST00000376387.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007843.hg.1 |
ENST00000376387.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007843.hg.1 |
ENST00000376387.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007843.hg.1 |
ENST00000376387.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007843.hg.1 |
ENST00000376387.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007843.hg.1 |
ENST00000376387.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007843.hg.1 |
ENST00000376387.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007843.hg.1 |
ENST00000376387.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007843.hg.1 |
ENST00000376387.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007843.hg.1 |
ENST00000376387.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007843.hg.1 |
ENST00000376387.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007843.hg.1 |
ENST00000376387.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007843.hg.1 |
ENST00000376387.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007843.hg.1 |
ENST00000376387.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007843.hg.1 |
ENST00000376387.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007843.hg.1 |
ENST00000376387.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007843.hg.1 |
ENST00000376387.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007843.hg.1 |
ENST00000376387.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007843.hg.1 |
ENST00000376387.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007843.hg.1 |
ENST00000376387.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007843.hg.1 |
ENST00000376387.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007843.hg.1 |
ENST00000376387.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007843.hg.1 |
ENST00000376387.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007843.hg.1 |
ENST00000463709.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007843.hg.1 |
ENST00000463709.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007843.hg.1 |
ENST00000463709.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007843.hg.1 |
ENST00000463709.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007843.hg.1 |
ENST00000463709.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007843.hg.1 |
ENST00000463709.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007843.hg.1 |
ENST00000463709.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007843.hg.1 |
ENST00000466555.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007843.hg.1 |
ENST00000466555.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007843.hg.1 |
ENST00000466555.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007843.hg.1 |
ENST00000466555.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007843.hg.1 |
NM_004800.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007843.hg.1 |
NM_004800.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007843.hg.1 |
NM_004800.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007843.hg.1 |
NM_004800.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007843.hg.1 |
NM_004800.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007843.hg.1 |
NM_004800.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007843.hg.1 |
NM_004800.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007843.hg.1 |
NM_004800.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007843.hg.1 |
NM_004800.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007843.hg.1 |
NM_004800.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007843.hg.1 |
NM_004800.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007843.hg.1 |
NM_004800.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007843.hg.1 |
NM_004800.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007843.hg.1 |
NM_004800.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007843.hg.1 |
NM_004800.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007843.hg.1 |
NM_004800.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007843.hg.1 |
NM_004800.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007843.hg.1 |
NM_004800.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007843.hg.1 |
NM_004800.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007843.hg.1 |
NM_004800.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007843.hg.1 |
NM_004800.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007843.hg.1 |
NM_004800.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007843.hg.1 |
NM_004800.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007843.hg.1 |
NM_004800.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007843.hg.1 |
NM_004800.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13003768.hg.1 |
ENST00000376387.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13003768.hg.1 |
NM_004800.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13003768.hg.1 |
TC13000356.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR13003769.hg.1 |
ENST00000376387.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13003769.hg.1 |
NM_004800.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13003769.hg.1 |
TC13000356.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR13003770.hg.1 |
ENST00000376387.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13003770.hg.1 |
NM_004800.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13003770.hg.1 |
TC13000356.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1300007843.hg.1 |
ENST00000376387.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007843.hg.1 |
ENST00000376387.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007843.hg.1 |
ENST00000376387.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007843.hg.1 |
ENST00000376387.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007843.hg.1 |
ENST00000376387.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007843.hg.1 |
ENST00000376387.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007843.hg.1 |
ENST00000376387.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007843.hg.1 |
ENST00000376387.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007843.hg.1 |
ENST00000376387.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007843.hg.1 |
ENST00000376387.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007843.hg.1 |
ENST00000376387.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007843.hg.1 |
ENST00000376387.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007843.hg.1 |
ENST00000376387.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007843.hg.1 |
ENST00000376387.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007843.hg.1 |
ENST00000376387.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007843.hg.1 |
ENST00000376387.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007843.hg.1 |
ENST00000376387.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007843.hg.1 |
ENST00000376387.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007843.hg.1 |
ENST00000376387.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007843.hg.1 |
ENST00000376387.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007843.hg.1 |
ENST00000376387.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007843.hg.1 |
ENST00000376387.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007843.hg.1 |
ENST00000376387.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007843.hg.1 |
ENST00000463709.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007843.hg.1 |
ENST00000463709.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007843.hg.1 |
ENST00000463709.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007843.hg.1 |
ENST00000463709.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007843.hg.1 |
ENST00000463709.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007843.hg.1 |
ENST00000463709.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007843.hg.1 |
ENST00000463709.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007843.hg.1 |
ENST00000466555.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007843.hg.1 |
ENST00000466555.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007843.hg.1 |
ENST00000466555.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007843.hg.1 |
ENST00000466555.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007843.hg.1 |
NM_004800.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007843.hg.1 |
NM_004800.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007843.hg.1 |
NM_004800.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007843.hg.1 |
NM_004800.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007843.hg.1 |
NM_004800.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007843.hg.1 |
NM_004800.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007843.hg.1 |
NM_004800.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007843.hg.1 |
NM_004800.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007843.hg.1 |
NM_004800.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007843.hg.1 |
NM_004800.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007843.hg.1 |
NM_004800.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007843.hg.1 |
NM_004800.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007843.hg.1 |
NM_004800.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007843.hg.1 |
NM_004800.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007843.hg.1 |
NM_004800.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007843.hg.1 |
NM_004800.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007843.hg.1 |
NM_004800.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007843.hg.1 |
NM_004800.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007843.hg.1 |
NM_004800.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007843.hg.1 |
NM_004800.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007843.hg.1 |
NM_004800.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007843.hg.1 |
NM_004800.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007843.hg.1 |
NM_004800.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007843.hg.1 |
NM_004800.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007843.hg.1 |
NM_004800.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300152790.hg.1 |
ENST00000376387.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300152790.hg.1 |
NM_004800.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300152790.hg.1 |
ENST00000376387.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300152790.hg.1 |
NM_004800.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300152790.hg.1 |
TC1300007843.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1300152789.hg.1 |
ENST00000376387.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300152789.hg.1 |
NM_004800.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300152789.hg.1 |
ENST00000376387.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300152789.hg.1 |
NM_004800.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300152789.hg.1 |
TC1300007843.hg.1 |
Clariom D TC |