Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR20008778.hg.1 |
ENST00000217289.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20008778.hg.1 |
ENST00000339538.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20008778.hg.1 |
ENST00000536936.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20008778.hg.1 |
NM_017671.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20008778.hg.1 |
TC20000603.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000217289.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000217289.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000217289.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000217289.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000217289.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000217289.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000217289.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000217289.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000217289.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000217289.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000217289.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000217289.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000217289.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000217289.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000217289.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000217289.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000217289.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000217289.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000217289.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000217289.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000378844.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000378844.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000378844.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000378844.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000478194.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000478194.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000478194.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000478194.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000478194.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000478194.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000478194.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000478194.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000478194.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000536936.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000536936.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000536936.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000536936.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000536936.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000536936.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000536936.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000536936.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000536936.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000536936.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000536936.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000536936.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000536936.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000536936.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000536936.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000536936.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
NM_017671.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
NM_017671.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
NM_017671.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
NM_017671.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
NM_017671.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
NM_017671.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
NM_017671.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
NM_017671.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
NM_017671.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
NM_017671.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
NM_017671.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
NM_017671.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
NM_017671.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
NM_017671.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
NM_017671.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
NM_017671.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
NM_017671.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
NM_017671.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
NM_017671.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
NM_017671.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000158633.hg.1 |
ENST00000217289.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000158633.hg.1 |
ENST00000536936.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000158633.hg.1 |
NM_017671.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000158633.hg.1 |
TC2000008318.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000158633.hg.1 |
ENST00000217289.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000158633.hg.1 |
ENST00000536936.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000158633.hg.1 |
NM_017671.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000217289.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000217289.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000217289.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000217289.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000217289.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000217289.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000217289.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000217289.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000217289.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000217289.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000217289.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000217289.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000217289.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000217289.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000217289.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000217289.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000217289.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000217289.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000217289.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000217289.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000378844.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000378844.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000378844.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000378844.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000478194.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000478194.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000478194.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000478194.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000478194.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000478194.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000478194.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000478194.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000478194.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000536936.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000536936.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000536936.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000536936.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000536936.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000536936.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000536936.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000536936.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000536936.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000536936.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000536936.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000536936.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000536936.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000536936.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000536936.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000536936.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
NM_017671.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
NM_017671.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
NM_017671.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
NM_017671.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
NM_017671.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
NM_017671.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
NM_017671.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
NM_017671.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
NM_017671.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
NM_017671.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
NM_017671.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
NM_017671.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
NM_017671.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
NM_017671.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
NM_017671.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
NM_017671.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
NM_017671.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
NM_017671.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
NM_017671.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
NM_017671.4 |
Refseq |