Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013409.hg.1 |
ENST00000361720.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013409.hg.1 |
ENST00000361720.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013409.hg.1 |
ENST00000361720.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013409.hg.1 |
ENST00000361720.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013409.hg.1 |
ENST00000361720.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013409.hg.1 |
ENST00000361720.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013409.hg.1 |
ENST00000361720.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013409.hg.1 |
ENST00000361720.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013409.hg.1 |
ENST00000361720.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013409.hg.1 |
ENST00000361720.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013409.hg.1 |
ENST00000445019.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013409.hg.1 |
ENST00000445019.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013409.hg.1 |
ENST00000445019.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013409.hg.1 |
ENST00000450844.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013409.hg.1 |
ENST00000450844.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013409.hg.1 |
ENST00000450844.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013409.hg.1 |
ENST00000450844.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013409.hg.1 |
NM_022078.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013409.hg.1 |
NM_022078.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013409.hg.1 |
NM_022078.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013409.hg.1 |
NM_022078.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013409.hg.1 |
NM_022078.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013409.hg.1 |
NM_022078.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013409.hg.1 |
NM_022078.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013409.hg.1 |
NM_022078.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013409.hg.1 |
NM_022078.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100013409.hg.1 |
NM_022078.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01036987.hg.1 |
TC01002396.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01036987.hg.1 |
ENST00000361720.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01036987.hg.1 |
ENST00000536641.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01036987.hg.1 |
NM_022078.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013409.hg.1 |
ENST00000361720.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013409.hg.1 |
ENST00000361720.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013409.hg.1 |
ENST00000361720.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013409.hg.1 |
ENST00000361720.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013409.hg.1 |
ENST00000361720.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013409.hg.1 |
ENST00000361720.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013409.hg.1 |
ENST00000361720.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013409.hg.1 |
ENST00000361720.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013409.hg.1 |
ENST00000361720.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013409.hg.1 |
ENST00000361720.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013409.hg.1 |
ENST00000445019.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013409.hg.1 |
ENST00000445019.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013409.hg.1 |
ENST00000445019.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013409.hg.1 |
ENST00000450844.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013409.hg.1 |
ENST00000450844.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013409.hg.1 |
ENST00000450844.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013409.hg.1 |
ENST00000450844.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100013409.hg.1 |
NM_022078.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013409.hg.1 |
NM_022078.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013409.hg.1 |
NM_022078.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013409.hg.1 |
NM_022078.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013409.hg.1 |
NM_022078.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013409.hg.1 |
NM_022078.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013409.hg.1 |
NM_022078.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013409.hg.1 |
NM_022078.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013409.hg.1 |
NM_022078.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100013409.hg.1 |
NM_022078.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100199607.hg.1 |
ENST00000361720.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100199607.hg.1 |
ENST00000450844.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100199607.hg.1 |
NM_022078.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100199607.hg.1 |
ENST00000361720.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100199607.hg.1 |
ENST00000450844.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100199607.hg.1 |
NM_022078.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100199607.hg.1 |
TC0100013409.hg.1 |
Clariom D TC |