Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OSBPL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_015550.3 | NP_056365.1 | 17 - 18 | 2429 | 77 | |
NM_145320.2 | NP_663160.1 | 15 - 16 | 2009 | 77 | ||
NM_145321.2 | NP_663161.1 | 15 - 16 | 1994 | 77 | ||
NM_145322.2 | NP_663162.1 | 14 - 15 | 1901 | 77 | ||
NR_104111.1 | 16 - 17 | 2501 | 77 | |||
NR_104112.1 | 15 - 16 | 2408 | 77 | |||
XM_005249698.3 | XP_005249755.1 | 17 - 18 | 2149 | 77 | ||
XM_006715681.3 | XP_006715744.1 | 16 - 17 | 2323 | 77 | ||
XM_006715682.3 | XP_006715745.1 | 16 - 17 | 2327 | 77 | ||
XM_006715683.3 | XP_006715746.1 | 15 - 16 | 2234 | 77 | ||
XM_006715684.3 | XP_006715747.1 | 16 - 17 | 2086 | 77 | ||
XM_011515258.2 | XP_011513560.1 | 17 - 18 | 2226 | 77 | ||
XM_011515259.2 | XP_011513561.1 | 18 - 19 | 2723 | 77 | ||
XM_017011948.1 | XP_016867437.1 | 19 - 20 | 2586 | 77 | ||
XM_017011949.1 | XP_016867438.1 | 17 - 18 | 2615 | 77 | ||
GenBank mRNA | AB014604.2 | - | 17 - 18 | 2366 | 77 | |
AF392444.1 | - | 17 - 18 | 2338 | 77 | ||
AF491781.1 | - | 16 - 17 | 2056 | 77 | ||
AF491782.1 | - | 15 - 16 | 1963 | 77 | ||
AF491783.1 | - | 15 - 16 | 1948 | 77 | ||
AF491784.1 | - | 14 - 15 | 1855 | 77 | ||
AF491785.1 | - | 16 - 17 | 2455 | 77 | ||
AF491786.1 | - | 15 - 16 | 2362 | 77 | ||
AL832293.1 | - | 14 - 15 | 1665 | 77 | ||
AY008372.1 | - | 17 - 18 | 2259 | 77 | ||
BC017731.1 | - | 17 - 18 | 2358 | 3909164 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons |