Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007132.hg.1 |
ENST00000301908.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007132.hg.1 |
ENST00000301908.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007132.hg.1 |
ENST00000301908.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007132.hg.1 |
ENST00000301908.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007132.hg.1 |
ENST00000301908.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007132.hg.1 |
ENST00000301908.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007132.hg.1 |
ENST00000301908.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007132.hg.1 |
ENST00000301908.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007132.hg.1 |
ENST00000518479.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007132.hg.1 |
ENST00000518479.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007132.hg.1 |
ENST00000518479.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007132.hg.1 |
ENST00000518479.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007132.hg.1 |
ENST00000519592.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007132.hg.1 |
ENST00000519592.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007132.hg.1 |
ENST00000519592.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007132.hg.1 |
ENST00000519592.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007132.hg.1 |
ENST00000519592.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007132.hg.1 |
ENST00000519592.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007132.hg.1 |
ENST00000522209.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007132.hg.1 |
ENST00000522209.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007132.hg.1 |
ENST00000522209.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007132.hg.1 |
ENST00000522209.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007132.hg.1 |
ENST00000522209.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007132.hg.1 |
ENST00000522209.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007132.hg.1 |
NM_001284244.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007132.hg.1 |
NM_001284244.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007132.hg.1 |
NM_001284244.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007132.hg.1 |
NM_001284244.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007132.hg.1 |
NM_001284244.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007132.hg.1 |
NM_001284244.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007132.hg.1 |
NM_006228.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007132.hg.1 |
NM_006228.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007132.hg.1 |
NM_006228.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007132.hg.1 |
NM_006228.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007132.hg.1 |
NM_006228.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007132.hg.1 |
NM_006228.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007132.hg.1 |
NM_006228.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007132.hg.1 |
NM_006228.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08003163.hg.1 |
TC08000217.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08003163.hg.1 |
ENST00000301908.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08003163.hg.1 |
NM_006228.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08003165.hg.1 |
TC08000217.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08003165.hg.1 |
ENST00000301908.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08003165.hg.1 |
NM_006228.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08003164.hg.1 |
TC08000217.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08003164.hg.1 |
ENST00000301908.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08003164.hg.1 |
NM_006228.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007132.hg.1 |
ENST00000301908.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007132.hg.1 |
ENST00000301908.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007132.hg.1 |
ENST00000301908.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007132.hg.1 |
ENST00000301908.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007132.hg.1 |
ENST00000301908.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007132.hg.1 |
ENST00000301908.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007132.hg.1 |
ENST00000301908.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007132.hg.1 |
ENST00000301908.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007132.hg.1 |
ENST00000518479.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007132.hg.1 |
ENST00000518479.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007132.hg.1 |
ENST00000518479.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007132.hg.1 |
ENST00000518479.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007132.hg.1 |
ENST00000519592.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007132.hg.1 |
ENST00000519592.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007132.hg.1 |
ENST00000519592.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007132.hg.1 |
ENST00000519592.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007132.hg.1 |
ENST00000519592.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007132.hg.1 |
ENST00000519592.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007132.hg.1 |
ENST00000522209.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007132.hg.1 |
ENST00000522209.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007132.hg.1 |
ENST00000522209.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007132.hg.1 |
ENST00000522209.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007132.hg.1 |
ENST00000522209.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007132.hg.1 |
ENST00000522209.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007132.hg.1 |
NM_001284244.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007132.hg.1 |
NM_001284244.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007132.hg.1 |
NM_001284244.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007132.hg.1 |
NM_001284244.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007132.hg.1 |
NM_001284244.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007132.hg.1 |
NM_001284244.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007132.hg.1 |
NM_006228.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007132.hg.1 |
NM_006228.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007132.hg.1 |
NM_006228.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007132.hg.1 |
NM_006228.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007132.hg.1 |
NM_006228.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007132.hg.1 |
NM_006228.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007132.hg.1 |
NM_006228.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007132.hg.1 |
NM_006228.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800149017.hg.1 |
ENST00000301908.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800149017.hg.1 |
NM_006228.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800149017.hg.1 |
ENST00000301908.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800149017.hg.1 |
NM_006228.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800149017.hg.1 |
TC0800007132.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800149020.hg.1 |
ENST00000301908.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800149020.hg.1 |
ENST00000518479.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800149020.hg.1 |
NM_006228.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800149020.hg.1 |
ENST00000301908.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800149020.hg.1 |
ENST00000518479.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800149020.hg.1 |
NM_006228.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800149020.hg.1 |
TC0800007132.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800149018.hg.1 |
ENST00000301908.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800149018.hg.1 |
NM_006228.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800149018.hg.1 |
ENST00000301908.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800149018.hg.1 |
NM_006228.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800149018.hg.1 |
TC0800007132.hg.1 |
Clariom D TC |