Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008440.hg.1 |
ENST00000308249.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008440.hg.1 |
ENST00000308249.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008440.hg.1 |
ENST00000308249.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008440.hg.1 |
ENST00000308249.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008440.hg.1 |
ENST00000308249.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008440.hg.1 |
ENST00000308249.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008440.hg.1 |
ENST00000308249.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008440.hg.1 |
ENST00000308249.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008440.hg.1 |
ENST00000308249.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008440.hg.1 |
ENST00000308249.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008440.hg.1 |
ENST00000308249.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008440.hg.1 |
ENST00000308249.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008440.hg.1 |
NM_014906.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008440.hg.1 |
NM_014906.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008440.hg.1 |
NM_014906.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008440.hg.1 |
NM_014906.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008440.hg.1 |
NM_014906.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008440.hg.1 |
NM_014906.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008440.hg.1 |
NM_014906.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008440.hg.1 |
NM_014906.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008440.hg.1 |
NM_014906.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008440.hg.1 |
NM_014906.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008440.hg.1 |
NM_014906.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008440.hg.1 |
NM_014906.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008440.hg.1 |
NR_048561.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008440.hg.1 |
NR_048561.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008440.hg.1 |
NR_048561.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008440.hg.1 |
NR_048561.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008440.hg.1 |
NR_048561.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008440.hg.1 |
NR_048561.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008440.hg.1 |
NR_048561.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008440.hg.1 |
NR_048561.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008440.hg.1 |
NR_048561.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008440.hg.1 |
NR_048561.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008440.hg.1 |
NR_048561.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17009344.hg.1 |
ENST00000308249.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17009344.hg.1 |
NM_014906.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17009344.hg.1 |
TC17000715.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR17009342.hg.1 |
ENST00000308249.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17009342.hg.1 |
NM_014906.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17009342.hg.1 |
TC17000715.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR17009346.hg.1 |
ENST00000308249.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17009346.hg.1 |
ENST00000443121.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17009346.hg.1 |
NM_014906.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17009346.hg.1 |
TC17000715.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR17009341.hg.1 |
ENST00000308249.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17009341.hg.1 |
NM_014906.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17009341.hg.1 |
TC17000715.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1700162810.hg.1 |
ENST00000308249.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700162810.hg.1 |
NM_014906.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700162810.hg.1 |
NR_048561.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700162810.hg.1 |
ENST00000308249.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700162810.hg.1 |
NM_014906.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700162810.hg.1 |
NR_048561.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700162810.hg.1 |
TC1700008440.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700162809.hg.1 |
ENST00000308249.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700162809.hg.1 |
NM_014906.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700162809.hg.1 |
NR_048561.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700162809.hg.1 |
ENST00000308249.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700162809.hg.1 |
NM_014906.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700162809.hg.1 |
NR_048561.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700162809.hg.1 |
TC1700008440.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700162812.hg.1 |
ENST00000308249.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700162812.hg.1 |
NM_014906.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700162812.hg.1 |
NR_048561.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700162812.hg.1 |
ENST00000308249.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700162812.hg.1 |
NM_014906.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700162812.hg.1 |
NR_048561.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700162812.hg.1 |
TC1700008440.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1700008440.hg.1 |
ENST00000308249.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008440.hg.1 |
ENST00000308249.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008440.hg.1 |
ENST00000308249.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008440.hg.1 |
ENST00000308249.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008440.hg.1 |
ENST00000308249.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008440.hg.1 |
ENST00000308249.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008440.hg.1 |
ENST00000308249.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008440.hg.1 |
ENST00000308249.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008440.hg.1 |
ENST00000308249.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008440.hg.1 |
ENST00000308249.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008440.hg.1 |
ENST00000308249.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008440.hg.1 |
ENST00000308249.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008440.hg.1 |
NM_014906.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008440.hg.1 |
NM_014906.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008440.hg.1 |
NM_014906.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008440.hg.1 |
NM_014906.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008440.hg.1 |
NM_014906.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008440.hg.1 |
NM_014906.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008440.hg.1 |
NM_014906.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008440.hg.1 |
NM_014906.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008440.hg.1 |
NM_014906.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008440.hg.1 |
NM_014906.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008440.hg.1 |
NM_014906.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008440.hg.1 |
NM_014906.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008440.hg.1 |
NR_048561.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008440.hg.1 |
NR_048561.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008440.hg.1 |
NR_048561.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008440.hg.1 |
NR_048561.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008440.hg.1 |
NR_048561.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008440.hg.1 |
NR_048561.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008440.hg.1 |
NR_048561.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008440.hg.1 |
NR_048561.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008440.hg.1 |
NR_048561.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008440.hg.1 |
NR_048561.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008440.hg.1 |
NR_048561.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700162811.hg.1 |
ENST00000308249.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700162811.hg.1 |
NM_014906.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700162811.hg.1 |
NR_048561.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700162811.hg.1 |
ENST00000308249.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700162811.hg.1 |
NM_014906.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700162811.hg.1 |
NR_048561.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700162811.hg.1 |
TC1700008440.hg.1 |
Clariom D TC |