Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X005597.hg.1 |
TC0X000424.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X005597.hg.1 |
ENST00000341514.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X005597.hg.1 |
ENST00000343533.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X005597.hg.1 |
ENST00000355691.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X005597.hg.1 |
ENST00000400860.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X005597.hg.1 |
NM_000052.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00152761.hg.1 |
ENST00000341514.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00152761.hg.1 |
ENST00000343533.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00152761.hg.1 |
ENST00000350425.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00152761.hg.1 |
NM_000052.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00152761.hg.1 |
NM_001282224.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00152761.hg.1 |
ENST00000341514.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00152761.hg.1 |
ENST00000343533.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00152761.hg.1 |
ENST00000350425.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00152761.hg.1 |
NM_000052.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00152761.hg.1 |
NM_001282224.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00152761.hg.1 |
TC0X00007707.hg.1 |
Clariom D TC |