Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006707.hg.1 |
ENST00000346208.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006707.hg.1 |
ENST00000346208.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006707.hg.1 |
ENST00000346208.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006707.hg.1 |
ENST00000346208.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006707.hg.1 |
ENST00000346208.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006707.hg.1 |
ENST00000346208.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006707.hg.1 |
ENST00000346208.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006707.hg.1 |
ENST00000346208.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006707.hg.1 |
ENST00000346208.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006707.hg.1 |
ENST00000346208.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006707.hg.1 |
ENST00000346208.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006707.hg.1 |
ENST00000346208.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006707.hg.1 |
ENST00000379328.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006707.hg.1 |
ENST00000379328.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006707.hg.1 |
ENST00000379328.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006707.hg.1 |
ENST00000379328.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006707.hg.1 |
ENST00000379328.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006707.hg.1 |
ENST00000379328.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006707.hg.1 |
ENST00000379328.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006707.hg.1 |
ENST00000379328.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006707.hg.1 |
ENST00000379328.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006707.hg.1 |
ENST00000379328.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006707.hg.1 |
ENST00000379328.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006707.hg.1 |
ENST00000379328.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006707.hg.1 |
ENST00000379328.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006707.hg.1 |
ENST00000379328.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006707.hg.1 |
ENST00000461472.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006707.hg.1 |
ENST00000461472.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006707.hg.1 |
ENST00000461472.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006707.hg.1 |
ENST00000461472.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006707.hg.1 |
ENST00000481743.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006707.hg.1 |
ENST00000481743.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006707.hg.1 |
NM_001002295.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006707.hg.1 |
NM_001002295.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006707.hg.1 |
NM_001002295.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006707.hg.1 |
NM_001002295.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006707.hg.1 |
NM_001002295.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006707.hg.1 |
NM_001002295.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006707.hg.1 |
NM_001002295.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006707.hg.1 |
NM_001002295.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006707.hg.1 |
NM_001002295.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006707.hg.1 |
NM_001002295.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006707.hg.1 |
NM_001002295.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006707.hg.1 |
NM_001002295.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006707.hg.1 |
NM_001002295.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006707.hg.1 |
NM_001002295.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006707.hg.1 |
NM_002051.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006707.hg.1 |
NM_002051.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006707.hg.1 |
NM_002051.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006707.hg.1 |
NM_002051.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006707.hg.1 |
NM_002051.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006707.hg.1 |
NM_002051.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006707.hg.1 |
NM_002051.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006707.hg.1 |
NM_002051.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006707.hg.1 |
NM_002051.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006707.hg.1 |
NM_002051.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006707.hg.1 |
NM_002051.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006707.hg.1 |
NM_002051.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006707.hg.1 |
NM_002051.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006707.hg.1 |
NM_002051.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10000799.hg.1 |
ENST00000346208.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10000799.hg.1 |
ENST00000379328.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10000799.hg.1 |
NM_001002295.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10000799.hg.1 |
NM_002051.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10000799.hg.1 |
TC10000070.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1000006707.hg.1 |
ENST00000346208.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006707.hg.1 |
ENST00000346208.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006707.hg.1 |
ENST00000346208.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006707.hg.1 |
ENST00000346208.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006707.hg.1 |
ENST00000346208.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006707.hg.1 |
ENST00000346208.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006707.hg.1 |
ENST00000346208.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006707.hg.1 |
ENST00000346208.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006707.hg.1 |
ENST00000346208.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006707.hg.1 |
ENST00000346208.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006707.hg.1 |
ENST00000346208.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006707.hg.1 |
ENST00000346208.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006707.hg.1 |
ENST00000379328.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006707.hg.1 |
ENST00000379328.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006707.hg.1 |
ENST00000379328.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006707.hg.1 |
ENST00000379328.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006707.hg.1 |
ENST00000379328.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006707.hg.1 |
ENST00000379328.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006707.hg.1 |
ENST00000379328.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006707.hg.1 |
ENST00000379328.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006707.hg.1 |
ENST00000379328.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006707.hg.1 |
ENST00000379328.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006707.hg.1 |
ENST00000379328.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006707.hg.1 |
ENST00000379328.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006707.hg.1 |
ENST00000379328.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006707.hg.1 |
ENST00000379328.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006707.hg.1 |
ENST00000461472.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006707.hg.1 |
ENST00000461472.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006707.hg.1 |
ENST00000461472.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006707.hg.1 |
ENST00000461472.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006707.hg.1 |
ENST00000481743.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006707.hg.1 |
ENST00000481743.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006707.hg.1 |
NM_001002295.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006707.hg.1 |
NM_001002295.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006707.hg.1 |
NM_001002295.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006707.hg.1 |
NM_001002295.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006707.hg.1 |
NM_001002295.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006707.hg.1 |
NM_001002295.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006707.hg.1 |
NM_001002295.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006707.hg.1 |
NM_001002295.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006707.hg.1 |
NM_001002295.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006707.hg.1 |
NM_001002295.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006707.hg.1 |
NM_001002295.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006707.hg.1 |
NM_001002295.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006707.hg.1 |
NM_001002295.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006707.hg.1 |
NM_001002295.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006707.hg.1 |
NM_002051.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006707.hg.1 |
NM_002051.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006707.hg.1 |
NM_002051.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006707.hg.1 |
NM_002051.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006707.hg.1 |
NM_002051.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006707.hg.1 |
NM_002051.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006707.hg.1 |
NM_002051.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006707.hg.1 |
NM_002051.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006707.hg.1 |
NM_002051.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006707.hg.1 |
NM_002051.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006707.hg.1 |
NM_002051.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006707.hg.1 |
NM_002051.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006707.hg.1 |
NM_002051.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006707.hg.1 |
NM_002051.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000146032.hg.1 |
ENST00000346208.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000146032.hg.1 |
ENST00000379328.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000146032.hg.1 |
NM_001002295.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000146032.hg.1 |
NM_002051.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000146032.hg.1 |
ENST00000346208.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000146032.hg.1 |
ENST00000379328.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000146032.hg.1 |
NM_001002295.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000146032.hg.1 |
NM_002051.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000146032.hg.1 |
TC1000006707.hg.1 |
Clariom D TC |