Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR22003040.hg.1 |
ENST00000215855.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22003040.hg.1 |
ENST00000404334.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22003040.hg.1 |
NM_004076.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22003040.hg.1 |
TC22000160.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR22003042.hg.1 |
ENST00000215855.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22003042.hg.1 |
ENST00000404334.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22003042.hg.1 |
NM_004076.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22003042.hg.1 |
TC22000160.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR22003038.hg.1 |
ENST00000215855.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22003038.hg.1 |
NM_004076.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22003038.hg.1 |
TC22000160.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006912.hg.1 |
ENST00000215855.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006912.hg.1 |
ENST00000215855.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006912.hg.1 |
ENST00000215855.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006912.hg.1 |
ENST00000215855.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006912.hg.1 |
ENST00000215855.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006912.hg.1 |
ENST00000215855.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006912.hg.1 |
ENST00000215855.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006912.hg.1 |
ENST00000404334.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006912.hg.1 |
ENST00000404334.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006912.hg.1 |
ENST00000404334.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006912.hg.1 |
ENST00000404334.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006912.hg.1 |
ENST00000404334.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006912.hg.1 |
NM_004076.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006912.hg.1 |
NM_004076.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006912.hg.1 |
NM_004076.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006912.hg.1 |
NM_004076.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006912.hg.1 |
NM_004076.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006912.hg.1 |
NM_004076.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200006912.hg.1 |
NM_004076.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22003039.hg.1 |
ENST00000215855.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22003039.hg.1 |
ENST00000404334.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22003039.hg.1 |
NM_004076.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22003039.hg.1 |
TC22000160.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC2200006912.hg.1 |
ENST00000215855.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006912.hg.1 |
ENST00000215855.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006912.hg.1 |
ENST00000215855.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006912.hg.1 |
ENST00000215855.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006912.hg.1 |
ENST00000215855.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006912.hg.1 |
ENST00000215855.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006912.hg.1 |
ENST00000215855.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006912.hg.1 |
ENST00000404334.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006912.hg.1 |
ENST00000404334.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006912.hg.1 |
ENST00000404334.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006912.hg.1 |
ENST00000404334.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006912.hg.1 |
ENST00000404334.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200006912.hg.1 |
NM_004076.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200006912.hg.1 |
NM_004076.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200006912.hg.1 |
NM_004076.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200006912.hg.1 |
NM_004076.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200006912.hg.1 |
NM_004076.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200006912.hg.1 |
NM_004076.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200006912.hg.1 |
NM_004076.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200147993.hg.1 |
TC2200006912.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200147993.hg.1 |
ENST00000215855.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200147993.hg.1 |
ENST00000404334.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200147993.hg.1 |
NM_004076.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200147993.hg.1 |
ENST00000215855.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200147993.hg.1 |
ENST00000404334.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200147993.hg.1 |
NM_004076.4 |
Refseq |