Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007777.hg.1 |
ENST00000373173.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007777.hg.1 |
ENST00000373173.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007777.hg.1 |
ENST00000373173.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007777.hg.1 |
ENST00000373173.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007777.hg.1 |
ENST00000373173.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007777.hg.1 |
ENST00000373173.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007777.hg.1 |
ENST00000373173.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007777.hg.1 |
ENST00000373173.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007777.hg.1 |
ENST00000373173.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007777.hg.1 |
ENST00000373173.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007777.hg.1 |
ENST00000373173.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007777.hg.1 |
ENST00000613473.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007777.hg.1 |
ENST00000613473.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007777.hg.1 |
ENST00000613473.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007777.hg.1 |
ENST00000613473.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007777.hg.1 |
ENST00000613473.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007777.hg.1 |
ENST00000613473.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007777.hg.1 |
ENST00000613473.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007777.hg.1 |
ENST00000613473.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007777.hg.1 |
ENST00000613473.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007777.hg.1 |
ENST00000613473.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007777.hg.1 |
ENST00000613473.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007777.hg.1 |
ENST00000622540.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007777.hg.1 |
ENST00000622540.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007777.hg.1 |
ENST00000622540.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007777.hg.1 |
ENST00000622540.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007777.hg.1 |
ENST00000622540.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007777.hg.1 |
ENST00000622540.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007777.hg.1 |
ENST00000622540.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007777.hg.1 |
ENST00000622540.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007777.hg.1 |
ENST00000622540.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007777.hg.1 |
ENST00000622540.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007777.hg.1 |
ENST00000622540.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007777.hg.1 |
NM_198450.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007777.hg.1 |
NM_198450.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007777.hg.1 |
NM_198450.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007777.hg.1 |
NM_198450.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007777.hg.1 |
NM_198450.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007777.hg.1 |
NM_198450.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007777.hg.1 |
NM_198450.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007777.hg.1 |
NM_198450.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007777.hg.1 |
NM_198450.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007777.hg.1 |
NM_198450.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007777.hg.1 |
NM_198450.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X005775.hg.1 |
TC0X000439.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X005775.hg.1 |
ENST00000373169.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X005775.hg.1 |
ENST00000373173.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X005775.hg.1 |
NM_198450.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007777.hg.1 |
ENST00000373173.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007777.hg.1 |
ENST00000373173.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007777.hg.1 |
ENST00000373173.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007777.hg.1 |
ENST00000373173.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007777.hg.1 |
ENST00000373173.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007777.hg.1 |
ENST00000373173.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007777.hg.1 |
ENST00000373173.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007777.hg.1 |
ENST00000373173.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007777.hg.1 |
ENST00000373173.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007777.hg.1 |
ENST00000373173.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007777.hg.1 |
ENST00000373173.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007777.hg.1 |
ENST00000613473.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007777.hg.1 |
ENST00000613473.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007777.hg.1 |
ENST00000613473.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007777.hg.1 |
ENST00000613473.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007777.hg.1 |
ENST00000613473.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007777.hg.1 |
ENST00000613473.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007777.hg.1 |
ENST00000613473.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007777.hg.1 |
ENST00000613473.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007777.hg.1 |
ENST00000613473.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007777.hg.1 |
ENST00000613473.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007777.hg.1 |
ENST00000613473.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007777.hg.1 |
ENST00000622540.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007777.hg.1 |
ENST00000622540.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007777.hg.1 |
ENST00000622540.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007777.hg.1 |
ENST00000622540.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007777.hg.1 |
ENST00000622540.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007777.hg.1 |
ENST00000622540.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007777.hg.1 |
ENST00000622540.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007777.hg.1 |
ENST00000622540.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007777.hg.1 |
ENST00000622540.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007777.hg.1 |
ENST00000622540.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007777.hg.1 |
ENST00000622540.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007777.hg.1 |
NM_198450.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007777.hg.1 |
NM_198450.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007777.hg.1 |
NM_198450.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007777.hg.1 |
NM_198450.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007777.hg.1 |
NM_198450.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007777.hg.1 |
NM_198450.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007777.hg.1 |
NM_198450.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007777.hg.1 |
NM_198450.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007777.hg.1 |
NM_198450.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007777.hg.1 |
NM_198450.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007777.hg.1 |
NM_198450.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00153136.hg.1 |
ENST00000373173.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00153136.hg.1 |
ENST00000613473.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00153136.hg.1 |
ENST00000622540.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00153136.hg.1 |
NM_198450.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00153136.hg.1 |
ENST00000373173.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00153136.hg.1 |
ENST00000613473.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00153136.hg.1 |
ENST00000622540.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00153136.hg.1 |
NM_198450.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00153136.hg.1 |
TC0X00007777.hg.1 |
Clariom D TC |