Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR10019107.hg.1 |
ENST00000371697.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10019107.hg.1 |
NM_014391.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10019107.hg.1 |
TC10001517.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011400.hg.1 |
ENST00000371697.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011400.hg.1 |
ENST00000371697.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011400.hg.1 |
ENST00000371697.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011400.hg.1 |
ENST00000371697.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011400.hg.1 |
ENST00000371697.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011400.hg.1 |
ENST00000371697.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011400.hg.1 |
ENST00000371697.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011400.hg.1 |
ENST00000371697.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011400.hg.1 |
ENST00000371697.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011400.hg.1 |
ENST00000371697.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011400.hg.1 |
ENST00000371697.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011400.hg.1 |
ENST00000371697.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011400.hg.1 |
NM_014391.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011400.hg.1 |
NM_014391.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011400.hg.1 |
NM_014391.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011400.hg.1 |
NM_014391.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011400.hg.1 |
NM_014391.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011400.hg.1 |
NM_014391.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011400.hg.1 |
NM_014391.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011400.hg.1 |
NM_014391.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011400.hg.1 |
NM_014391.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011400.hg.1 |
NM_014391.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011400.hg.1 |
NM_014391.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011400.hg.1 |
NM_014391.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000177438.hg.1 |
ENST00000371697.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000177438.hg.1 |
NM_014391.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000177438.hg.1 |
ENST00000371697.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000177438.hg.1 |
NM_014391.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000177438.hg.1 |
TC1000011400.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1000011400.hg.1 |
ENST00000371697.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011400.hg.1 |
ENST00000371697.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011400.hg.1 |
ENST00000371697.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011400.hg.1 |
ENST00000371697.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011400.hg.1 |
ENST00000371697.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011400.hg.1 |
ENST00000371697.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011400.hg.1 |
ENST00000371697.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011400.hg.1 |
ENST00000371697.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011400.hg.1 |
ENST00000371697.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011400.hg.1 |
ENST00000371697.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011400.hg.1 |
ENST00000371697.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011400.hg.1 |
ENST00000371697.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011400.hg.1 |
NM_014391.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011400.hg.1 |
NM_014391.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011400.hg.1 |
NM_014391.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011400.hg.1 |
NM_014391.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011400.hg.1 |
NM_014391.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011400.hg.1 |
NM_014391.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011400.hg.1 |
NM_014391.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011400.hg.1 |
NM_014391.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011400.hg.1 |
NM_014391.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011400.hg.1 |
NM_014391.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011400.hg.1 |
NM_014391.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011400.hg.1 |
NM_014391.2 |
Refseq |