Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012485.hg.1 |
ENST00000199814.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012485.hg.1 |
ENST00000199814.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012485.hg.1 |
ENST00000199814.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012485.hg.1 |
ENST00000199814.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012485.hg.1 |
ENST00000199814.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012485.hg.1 |
ENST00000199814.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012485.hg.1 |
ENST00000199814.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012485.hg.1 |
ENST00000199814.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012485.hg.1 |
ENST00000199814.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012485.hg.1 |
ENST00000199814.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012485.hg.1 |
ENST00000199814.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012485.hg.1 |
ENST00000199814.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012485.hg.1 |
ENST00000199814.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012485.hg.1 |
ENST00000199814.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012485.hg.1 |
ENST00000199814.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012485.hg.1 |
ENST00000199814.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012485.hg.1 |
ENST00000199814.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012485.hg.1 |
ENST00000447771.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012485.hg.1 |
ENST00000447771.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012485.hg.1 |
ENST00000447771.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012485.hg.1 |
ENST00000447771.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012485.hg.1 |
ENST00000447771.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012485.hg.1 |
ENST00000447771.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012485.hg.1 |
ENST00000447771.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012485.hg.1 |
ENST00000447771.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012485.hg.1 |
ENST00000447771.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012485.hg.1 |
ENST00000447771.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012485.hg.1 |
ENST00000447771.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012485.hg.1 |
ENST00000447771.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012485.hg.1 |
ENST00000447771.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012485.hg.1 |
ENST00000518917.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012485.hg.1 |
ENST00000518917.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012485.hg.1 |
ENST00000518917.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012485.hg.1 |
ENST00000518917.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012485.hg.1 |
ENST00000518917.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012485.hg.1 |
ENST00000518917.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012485.hg.1 |
ENST00000518917.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012485.hg.1 |
ENST00000518917.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012485.hg.1 |
ENST00000520132.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012485.hg.1 |
ENST00000520132.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012485.hg.1 |
ENST00000520132.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012485.hg.1 |
ENST00000520132.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012485.hg.1 |
ENST00000520132.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012485.hg.1 |
ENST00000520132.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012485.hg.1 |
ENST00000520132.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012485.hg.1 |
ENST00000521248.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012485.hg.1 |
ENST00000521248.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012485.hg.1 |
ENST00000521248.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012485.hg.1 |
ENST00000521248.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012485.hg.1 |
ENST00000521248.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012485.hg.1 |
ENST00000521248.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012485.hg.1 |
ENST00000521248.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012485.hg.1 |
ENST00000521248.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012485.hg.1 |
ENST00000521248.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012485.hg.1 |
ENST00000521464.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012485.hg.1 |
ENST00000521464.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012485.hg.1 |
ENST00000521464.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012485.hg.1 |
ENST00000521464.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012485.hg.1 |
ENST00000521464.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012485.hg.1 |
ENST00000521594.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012485.hg.1 |
ENST00000521594.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012485.hg.1 |
ENST00000521594.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012485.hg.1 |
ENST00000521594.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012485.hg.1 |
ENST00000521594.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012485.hg.1 |
ENST00000521594.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012485.hg.1 |
ENST00000522469.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012485.hg.1 |
ENST00000522469.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012485.hg.1 |
ENST00000522469.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012485.hg.1 |
ENST00000522469.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012485.hg.1 |
ENST00000522469.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012485.hg.1 |
ENST00000522469.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012485.hg.1 |
ENST00000522469.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012485.hg.1 |
ENST00000522469.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012485.hg.1 |
ENST00000522469.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012485.hg.1 |
ENST00000522469.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012485.hg.1 |
NM_018047.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012485.hg.1 |
NM_018047.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012485.hg.1 |
NM_018047.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012485.hg.1 |
NM_018047.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012485.hg.1 |
NM_018047.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012485.hg.1 |
NM_018047.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012485.hg.1 |
NM_018047.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012485.hg.1 |
NM_018047.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012485.hg.1 |
NM_018047.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012485.hg.1 |
NM_018047.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012485.hg.1 |
NM_018047.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012485.hg.1 |
NM_018047.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012485.hg.1 |
NM_018047.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012485.hg.1 |
NM_018047.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012485.hg.1 |
NM_018047.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012485.hg.1 |
NM_018047.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012485.hg.1 |
NM_018047.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05026906.hg.1 |
TC05001942.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05026906.hg.1 |
ENST00000199814.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05026906.hg.1 |
ENST00000447771.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05026906.hg.1 |
ENST00000540000.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05026906.hg.1 |
NM_018047.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500186968.hg.1 |
ENST00000199814.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500186968.hg.1 |
ENST00000447771.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500186968.hg.1 |
ENST00000518917.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500186968.hg.1 |
ENST00000521248.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500186968.hg.1 |
ENST00000521464.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500186968.hg.1 |
ENST00000521594.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500186968.hg.1 |
ENST00000522469.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500186968.hg.1 |
NM_018047.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500186968.hg.1 |
ENST00000199814.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500186968.hg.1 |
ENST00000447771.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500186968.hg.1 |
ENST00000518917.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500186968.hg.1 |
ENST00000521248.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500186968.hg.1 |
ENST00000521464.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500186968.hg.1 |
ENST00000521594.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500186968.hg.1 |
ENST00000522469.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500186968.hg.1 |
NM_018047.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500186968.hg.1 |
TC0500012485.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0500012485.hg.1 |
ENST00000199814.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012485.hg.1 |
ENST00000199814.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012485.hg.1 |
ENST00000199814.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012485.hg.1 |
ENST00000199814.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012485.hg.1 |
ENST00000199814.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012485.hg.1 |
ENST00000199814.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012485.hg.1 |
ENST00000199814.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012485.hg.1 |
ENST00000199814.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012485.hg.1 |
ENST00000199814.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012485.hg.1 |
ENST00000199814.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012485.hg.1 |
ENST00000199814.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012485.hg.1 |
ENST00000199814.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012485.hg.1 |
ENST00000199814.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012485.hg.1 |
ENST00000199814.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012485.hg.1 |
ENST00000199814.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012485.hg.1 |
ENST00000199814.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012485.hg.1 |
ENST00000199814.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012485.hg.1 |
ENST00000447771.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012485.hg.1 |
ENST00000447771.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012485.hg.1 |
ENST00000447771.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012485.hg.1 |
ENST00000447771.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012485.hg.1 |
ENST00000447771.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012485.hg.1 |
ENST00000447771.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012485.hg.1 |
ENST00000447771.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012485.hg.1 |
ENST00000447771.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012485.hg.1 |
ENST00000447771.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012485.hg.1 |
ENST00000447771.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012485.hg.1 |
ENST00000447771.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012485.hg.1 |
ENST00000447771.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012485.hg.1 |
ENST00000447771.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012485.hg.1 |
ENST00000518917.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012485.hg.1 |
ENST00000518917.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012485.hg.1 |
ENST00000518917.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012485.hg.1 |
ENST00000518917.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012485.hg.1 |
ENST00000518917.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012485.hg.1 |
ENST00000518917.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012485.hg.1 |
ENST00000518917.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012485.hg.1 |
ENST00000518917.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012485.hg.1 |
ENST00000520132.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012485.hg.1 |
ENST00000520132.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012485.hg.1 |
ENST00000520132.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012485.hg.1 |
ENST00000520132.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012485.hg.1 |
ENST00000520132.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012485.hg.1 |
ENST00000520132.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012485.hg.1 |
ENST00000520132.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012485.hg.1 |
ENST00000521248.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012485.hg.1 |
ENST00000521248.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012485.hg.1 |
ENST00000521248.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012485.hg.1 |
ENST00000521248.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012485.hg.1 |
ENST00000521248.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012485.hg.1 |
ENST00000521248.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012485.hg.1 |
ENST00000521248.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012485.hg.1 |
ENST00000521248.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012485.hg.1 |
ENST00000521248.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012485.hg.1 |
ENST00000521464.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012485.hg.1 |
ENST00000521464.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012485.hg.1 |
ENST00000521464.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012485.hg.1 |
ENST00000521464.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012485.hg.1 |
ENST00000521464.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012485.hg.1 |
ENST00000521594.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012485.hg.1 |
ENST00000521594.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012485.hg.1 |
ENST00000521594.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012485.hg.1 |
ENST00000521594.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012485.hg.1 |
ENST00000521594.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012485.hg.1 |
ENST00000521594.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012485.hg.1 |
ENST00000522469.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012485.hg.1 |
ENST00000522469.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012485.hg.1 |
ENST00000522469.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012485.hg.1 |
ENST00000522469.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012485.hg.1 |
ENST00000522469.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012485.hg.1 |
ENST00000522469.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012485.hg.1 |
ENST00000522469.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012485.hg.1 |
ENST00000522469.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012485.hg.1 |
ENST00000522469.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012485.hg.1 |
ENST00000522469.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012485.hg.1 |
NM_018047.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012485.hg.1 |
NM_018047.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012485.hg.1 |
NM_018047.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012485.hg.1 |
NM_018047.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012485.hg.1 |
NM_018047.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012485.hg.1 |
NM_018047.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012485.hg.1 |
NM_018047.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012485.hg.1 |
NM_018047.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012485.hg.1 |
NM_018047.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012485.hg.1 |
NM_018047.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012485.hg.1 |
NM_018047.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012485.hg.1 |
NM_018047.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012485.hg.1 |
NM_018047.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012485.hg.1 |
NM_018047.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012485.hg.1 |
NM_018047.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012485.hg.1 |
NM_018047.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012485.hg.1 |
NM_018047.2 |
Refseq |