Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR09020714.hg.1 |
TC09001629.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09020714.hg.1 |
ENST00000372890.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09020714.hg.1 |
ENST00000460320.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09020714.hg.1 |
ENST00000546104.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09020714.hg.1 |
NM_015679.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011634.hg.1 |
ENST00000372890.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011634.hg.1 |
ENST00000372890.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011634.hg.1 |
ENST00000372890.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011634.hg.1 |
ENST00000372890.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011634.hg.1 |
ENST00000372890.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011634.hg.1 |
ENST00000372890.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011634.hg.1 |
ENST00000372890.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011634.hg.1 |
ENST00000372890.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011634.hg.1 |
ENST00000372890.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011634.hg.1 |
ENST00000372890.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011634.hg.1 |
ENST00000372890.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011634.hg.1 |
ENST00000372890.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011634.hg.1 |
ENST00000372890.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011634.hg.1 |
ENST00000372890.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011634.hg.1 |
ENST00000372890.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011634.hg.1 |
ENST00000372890.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011634.hg.1 |
ENST00000372890.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011634.hg.1 |
ENST00000460320.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011634.hg.1 |
ENST00000460320.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011634.hg.1 |
ENST00000460320.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011634.hg.1 |
ENST00000460320.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011634.hg.1 |
ENST00000460320.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011634.hg.1 |
ENST00000460320.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011634.hg.1 |
ENST00000460320.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011634.hg.1 |
ENST00000460320.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011634.hg.1 |
ENST00000460320.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011634.hg.1 |
ENST00000460320.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011634.hg.1 |
ENST00000461180.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011634.hg.1 |
ENST00000461180.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011634.hg.1 |
ENST00000461180.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011634.hg.1 |
ENST00000608093.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011634.hg.1 |
ENST00000608252.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011634.hg.1 |
NM_015679.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011634.hg.1 |
NM_015679.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011634.hg.1 |
NM_015679.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011634.hg.1 |
NM_015679.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011634.hg.1 |
NM_015679.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011634.hg.1 |
NM_015679.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011634.hg.1 |
NM_015679.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011634.hg.1 |
NM_015679.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011634.hg.1 |
NM_015679.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011634.hg.1 |
NM_015679.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011634.hg.1 |
NM_015679.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011634.hg.1 |
NM_015679.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09020716.hg.1 |
TC09001629.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09020716.hg.1 |
ENST00000372890.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09020716.hg.1 |
ENST00000460320.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09020716.hg.1 |
NM_015679.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900179616.hg.1 |
ENST00000372890.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900179616.hg.1 |
ENST00000460320.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900179616.hg.1 |
NM_015679.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900179616.hg.1 |
ENST00000372890.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900179616.hg.1 |
ENST00000460320.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900179616.hg.1 |
NM_015679.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900179616.hg.1 |
TC0900011634.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0900011634.hg.1 |
ENST00000372890.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011634.hg.1 |
ENST00000372890.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011634.hg.1 |
ENST00000372890.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011634.hg.1 |
ENST00000372890.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011634.hg.1 |
ENST00000372890.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011634.hg.1 |
ENST00000372890.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011634.hg.1 |
ENST00000372890.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011634.hg.1 |
ENST00000372890.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011634.hg.1 |
ENST00000372890.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011634.hg.1 |
ENST00000372890.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011634.hg.1 |
ENST00000372890.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011634.hg.1 |
ENST00000372890.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011634.hg.1 |
ENST00000372890.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011634.hg.1 |
ENST00000372890.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011634.hg.1 |
ENST00000372890.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011634.hg.1 |
ENST00000372890.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011634.hg.1 |
ENST00000372890.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011634.hg.1 |
ENST00000460320.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011634.hg.1 |
ENST00000460320.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011634.hg.1 |
ENST00000460320.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011634.hg.1 |
ENST00000460320.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011634.hg.1 |
ENST00000460320.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011634.hg.1 |
ENST00000460320.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011634.hg.1 |
ENST00000460320.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011634.hg.1 |
ENST00000460320.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011634.hg.1 |
ENST00000460320.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011634.hg.1 |
ENST00000460320.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011634.hg.1 |
ENST00000461180.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011634.hg.1 |
ENST00000461180.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011634.hg.1 |
ENST00000461180.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011634.hg.1 |
ENST00000608093.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011634.hg.1 |
ENST00000608252.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011634.hg.1 |
NM_015679.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011634.hg.1 |
NM_015679.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011634.hg.1 |
NM_015679.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011634.hg.1 |
NM_015679.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011634.hg.1 |
NM_015679.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011634.hg.1 |
NM_015679.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011634.hg.1 |
NM_015679.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011634.hg.1 |
NM_015679.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011634.hg.1 |
NM_015679.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011634.hg.1 |
NM_015679.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011634.hg.1 |
NM_015679.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011634.hg.1 |
NM_015679.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900179615.hg.1 |
ENST00000372890.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900179615.hg.1 |
ENST00000460320.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900179615.hg.1 |
NM_015679.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900179615.hg.1 |
ENST00000372890.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900179615.hg.1 |
ENST00000460320.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900179615.hg.1 |
NM_015679.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900179615.hg.1 |
TC0900011634.hg.1 |
Clariom D TC |