Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR17001977.hg.1 |
ENST00000173229.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17001977.hg.1 |
ENST00000538852.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17001977.hg.1 |
ENST00000546090.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17001977.hg.1 |
NM_004822.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17001977.hg.1 |
TC17000143.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006839.hg.1 |
ENST00000173229.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006839.hg.1 |
ENST00000173229.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006839.hg.1 |
ENST00000173229.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006839.hg.1 |
ENST00000173229.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006839.hg.1 |
ENST00000173229.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006839.hg.1 |
ENST00000173229.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006839.hg.1 |
ENST00000173229.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006839.hg.1 |
ENST00000173229.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006839.hg.1 |
ENST00000173229.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006839.hg.1 |
ENST00000173229.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006839.hg.1 |
ENST00000436734.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006839.hg.1 |
ENST00000436734.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006839.hg.1 |
ENST00000436734.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006839.hg.1 |
ENST00000436734.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006839.hg.1 |
NM_004822.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006839.hg.1 |
NM_004822.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006839.hg.1 |
NM_004822.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006839.hg.1 |
NM_004822.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006839.hg.1 |
NM_004822.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006839.hg.1 |
NM_004822.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006839.hg.1 |
NM_004822.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006839.hg.1 |
NM_004822.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006839.hg.1 |
NM_004822.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006839.hg.1 |
NM_004822.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700148232.hg.1 |
ENST00000173229.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700148232.hg.1 |
NM_004822.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700148232.hg.1 |
ENST00000173229.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700148232.hg.1 |
NM_004822.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700148232.hg.1 |
TC1700006839.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1700006839.hg.1 |
ENST00000173229.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006839.hg.1 |
ENST00000173229.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006839.hg.1 |
ENST00000173229.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006839.hg.1 |
ENST00000173229.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006839.hg.1 |
ENST00000173229.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006839.hg.1 |
ENST00000173229.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006839.hg.1 |
ENST00000173229.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006839.hg.1 |
ENST00000173229.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006839.hg.1 |
ENST00000173229.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006839.hg.1 |
ENST00000173229.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006839.hg.1 |
ENST00000436734.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006839.hg.1 |
ENST00000436734.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006839.hg.1 |
ENST00000436734.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006839.hg.1 |
ENST00000436734.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006839.hg.1 |
NM_004822.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006839.hg.1 |
NM_004822.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006839.hg.1 |
NM_004822.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006839.hg.1 |
NM_004822.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006839.hg.1 |
NM_004822.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006839.hg.1 |
NM_004822.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006839.hg.1 |
NM_004822.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006839.hg.1 |
NM_004822.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006839.hg.1 |
NM_004822.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006839.hg.1 |
NM_004822.2 |
Refseq |