Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR09018942.hg.1 |
TC09001510.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09018942.hg.1 |
NM_001083885.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09018941.hg.1 |
TC09001510.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09018941.hg.1 |
ENST00000265134.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09018941.hg.1 |
NM_001083885.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011286.hg.1 |
ENST00000265134.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011286.hg.1 |
ENST00000265134.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011286.hg.1 |
ENST00000265134.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011286.hg.1 |
ENST00000265134.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011286.hg.1 |
ENST00000265134.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011286.hg.1 |
ENST00000265134.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011286.hg.1 |
ENST00000265134.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011286.hg.1 |
ENST00000265134.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011286.hg.1 |
ENST00000265134.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011286.hg.1 |
ENST00000265134.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011286.hg.1 |
ENST00000265134.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011286.hg.1 |
ENST00000265134.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011286.hg.1 |
ENST00000265134.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011286.hg.1 |
ENST00000265134.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011286.hg.1 |
ENST00000362057.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011286.hg.1 |
ENST00000362057.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011286.hg.1 |
ENST00000362057.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011286.hg.1 |
ENST00000362057.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011286.hg.1 |
ENST00000362057.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011286.hg.1 |
ENST00000362057.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011286.hg.1 |
ENST00000362057.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011286.hg.1 |
ENST00000362057.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011286.hg.1 |
ENST00000362057.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011286.hg.1 |
ENST00000362057.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011286.hg.1 |
ENST00000362057.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011286.hg.1 |
ENST00000362057.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011286.hg.1 |
ENST00000362057.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011286.hg.1 |
ENST00000362057.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011286.hg.1 |
ENST00000362057.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011286.hg.1 |
ENST00000362057.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011286.hg.1 |
ENST00000374057.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011286.hg.1 |
ENST00000374057.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011286.hg.1 |
ENST00000374057.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011286.hg.1 |
ENST00000374057.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011286.hg.1 |
ENST00000374057.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011286.hg.1 |
ENST00000374057.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011286.hg.1 |
ENST00000374057.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011286.hg.1 |
ENST00000374059.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011286.hg.1 |
ENST00000374059.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011286.hg.1 |
ENST00000374059.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011286.hg.1 |
ENST00000374059.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011286.hg.1 |
ENST00000374059.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011286.hg.1 |
ENST00000374059.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011286.hg.1 |
ENST00000374059.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011286.hg.1 |
ENST00000374059.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011286.hg.1 |
ENST00000374059.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011286.hg.1 |
ENST00000374059.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011286.hg.1 |
ENST00000374059.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011286.hg.1 |
ENST00000480518.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011286.hg.1 |
ENST00000480518.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011286.hg.1 |
NM_001083885.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011286.hg.1 |
NM_001083885.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011286.hg.1 |
NM_001083885.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011286.hg.1 |
NM_001083885.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011286.hg.1 |
NM_001083885.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011286.hg.1 |
NM_001083885.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011286.hg.1 |
NM_001083885.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011286.hg.1 |
NM_001083885.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011286.hg.1 |
NM_001083885.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011286.hg.1 |
NM_001083885.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011286.hg.1 |
NM_001083885.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011286.hg.1 |
NM_001083885.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011286.hg.1 |
NM_001083885.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011286.hg.1 |
NM_001083885.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011286.hg.1 |
NM_001083885.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011286.hg.1 |
NM_001173425.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011286.hg.1 |
NM_001173425.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011286.hg.1 |
NM_001173425.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011286.hg.1 |
NM_001173425.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011286.hg.1 |
NM_001173425.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011286.hg.1 |
NM_001173425.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011286.hg.1 |
NM_001173425.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011286.hg.1 |
NM_001173425.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011286.hg.1 |
NM_001173425.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011286.hg.1 |
NM_001173425.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011286.hg.1 |
NM_001173425.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011286.hg.1 |
NM_001173425.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011286.hg.1 |
NM_001173425.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011286.hg.1 |
NM_001173425.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011286.hg.1 |
NM_001173425.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011286.hg.1 |
NM_001173425.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011286.hg.1 |
NM_001173425.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011286.hg.1 |
NM_001173425.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011286.hg.1 |
NM_015404.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011286.hg.1 |
NM_015404.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011286.hg.1 |
NM_015404.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011286.hg.1 |
NM_015404.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011286.hg.1 |
NM_015404.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011286.hg.1 |
NM_015404.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011286.hg.1 |
NM_015404.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011286.hg.1 |
NM_015404.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011286.hg.1 |
NM_015404.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011286.hg.1 |
NM_015404.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011286.hg.1 |
NM_015404.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011286.hg.1 |
NM_015404.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011286.hg.1 |
NM_015404.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011286.hg.1 |
NM_015404.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011286.hg.1 |
NM_015404.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011286.hg.1 |
NM_015404.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011286.hg.1 |
NM_015404.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011286.hg.1 |
NM_015404.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900176972.hg.1 |
NM_001083885.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900176972.hg.1 |
NM_001083885.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900176972.hg.1 |
TC0900011286.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0900011286.hg.1 |
ENST00000265134.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011286.hg.1 |
ENST00000265134.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011286.hg.1 |
ENST00000265134.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011286.hg.1 |
ENST00000265134.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011286.hg.1 |
ENST00000265134.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011286.hg.1 |
ENST00000265134.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011286.hg.1 |
ENST00000265134.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011286.hg.1 |
ENST00000265134.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011286.hg.1 |
ENST00000265134.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011286.hg.1 |
ENST00000265134.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011286.hg.1 |
ENST00000265134.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011286.hg.1 |
ENST00000265134.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011286.hg.1 |
ENST00000265134.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011286.hg.1 |
ENST00000265134.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011286.hg.1 |
ENST00000362057.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011286.hg.1 |
ENST00000362057.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011286.hg.1 |
ENST00000362057.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011286.hg.1 |
ENST00000362057.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011286.hg.1 |
ENST00000362057.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011286.hg.1 |
ENST00000362057.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011286.hg.1 |
ENST00000362057.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011286.hg.1 |
ENST00000362057.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011286.hg.1 |
ENST00000362057.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011286.hg.1 |
ENST00000362057.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011286.hg.1 |
ENST00000362057.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011286.hg.1 |
ENST00000362057.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011286.hg.1 |
ENST00000362057.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011286.hg.1 |
ENST00000362057.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011286.hg.1 |
ENST00000362057.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011286.hg.1 |
ENST00000362057.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011286.hg.1 |
ENST00000374057.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011286.hg.1 |
ENST00000374057.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011286.hg.1 |
ENST00000374057.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011286.hg.1 |
ENST00000374057.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011286.hg.1 |
ENST00000374057.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011286.hg.1 |
ENST00000374057.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011286.hg.1 |
ENST00000374057.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011286.hg.1 |
ENST00000374059.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011286.hg.1 |
ENST00000374059.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011286.hg.1 |
ENST00000374059.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011286.hg.1 |
ENST00000374059.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011286.hg.1 |
ENST00000374059.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011286.hg.1 |
ENST00000374059.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011286.hg.1 |
ENST00000374059.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011286.hg.1 |
ENST00000374059.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011286.hg.1 |
ENST00000374059.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011286.hg.1 |
ENST00000374059.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011286.hg.1 |
ENST00000374059.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011286.hg.1 |
ENST00000480518.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011286.hg.1 |
ENST00000480518.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011286.hg.1 |
NM_001083885.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011286.hg.1 |
NM_001083885.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011286.hg.1 |
NM_001083885.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011286.hg.1 |
NM_001083885.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011286.hg.1 |
NM_001083885.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011286.hg.1 |
NM_001083885.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011286.hg.1 |
NM_001083885.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011286.hg.1 |
NM_001083885.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011286.hg.1 |
NM_001083885.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011286.hg.1 |
NM_001083885.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011286.hg.1 |
NM_001083885.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011286.hg.1 |
NM_001083885.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011286.hg.1 |
NM_001083885.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011286.hg.1 |
NM_001083885.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011286.hg.1 |
NM_001083885.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011286.hg.1 |
NM_001173425.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011286.hg.1 |
NM_001173425.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011286.hg.1 |
NM_001173425.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011286.hg.1 |
NM_001173425.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011286.hg.1 |
NM_001173425.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011286.hg.1 |
NM_001173425.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011286.hg.1 |
NM_001173425.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011286.hg.1 |
NM_001173425.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011286.hg.1 |
NM_001173425.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011286.hg.1 |
NM_001173425.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011286.hg.1 |
NM_001173425.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011286.hg.1 |
NM_001173425.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011286.hg.1 |
NM_001173425.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011286.hg.1 |
NM_001173425.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011286.hg.1 |
NM_001173425.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011286.hg.1 |
NM_001173425.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011286.hg.1 |
NM_001173425.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011286.hg.1 |
NM_001173425.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011286.hg.1 |
NM_015404.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011286.hg.1 |
NM_015404.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011286.hg.1 |
NM_015404.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011286.hg.1 |
NM_015404.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011286.hg.1 |
NM_015404.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011286.hg.1 |
NM_015404.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011286.hg.1 |
NM_015404.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011286.hg.1 |
NM_015404.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011286.hg.1 |
NM_015404.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011286.hg.1 |
NM_015404.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011286.hg.1 |
NM_015404.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011286.hg.1 |
NM_015404.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011286.hg.1 |
NM_015404.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011286.hg.1 |
NM_015404.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011286.hg.1 |
NM_015404.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011286.hg.1 |
NM_015404.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011286.hg.1 |
NM_015404.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011286.hg.1 |
NM_015404.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900176971.hg.1 |
ENST00000265134.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900176971.hg.1 |
NM_001083885.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900176971.hg.1 |
ENST00000265134.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900176971.hg.1 |
NM_001083885.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900176971.hg.1 |
TC0900011286.hg.1 |
Clariom D TC |